Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 33616651-33616918 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRYZL1
Entrez Gene ID
9946
Gene Name
crystallin zeta like 1
Gene Aliases
4P11, QOH-1
UniGene
Hs.352671
Gene Chromosome Location
Chr.21: 33589341-33641854 on Build GRCh38
Transcripts
NM_145858.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6581717 c.250G>A p.D84N Chr.21: 33616718-33616718 on Build GRCh38
6048874 c.221G>T p.G74V Chr.21: 33616747-33616747 on Build GRCh38
1632465 c.253G>A p.E85K Chr.21: 33616715-33616715 on Build GRCh38
6840132 c.262G>A p.G88R Chr.21: 33616706-33616706 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs996334717 Chr.21: 33616806-33616809 on Build GRCh38
rs890655756 Chr.21: 33616820-33616820 on Build GRCh38
rs756054203 Chr.21: 33616775-33616775 on Build GRCh38
rs1004614760 Chr.21: 33616821-33616822 on Build GRCh38
rs778140142 Chr.21: 33616739-33616739 on Build GRCh38
rs758963259 Chr.21: 33616714-33616716 on Build GRCh38
rs184229251 Chr.21: 33616839-33616839 on Build GRCh38
rs994915803 Chr.21: 33616845-33616845 on Build GRCh38
rs199987497 Chr.21: 33616719-33616719 on Build GRCh38
rs766525692 Chr.21: 33616679-33616679 on Build GRCh38
rs767644164 Chr.21: 33616869-33616870 on Build GRCh38
rs1034247944 Chr.21: 33616865-33616865 on Build GRCh38
rs963067348 Chr.21: 33616840-33616840 on Build GRCh38
rs1030517688 Chr.21: 33616808-33616808 on Build GRCh38
rs879056778 Chr.21: 33616785-33616786 on Build GRCh38
rs751793190 Chr.21: 33616686-33616686 on Build GRCh38
rs748218132 Chr.21: 33616772-33616772 on Build GRCh38
rs781328452 Chr.21: 33616721-33616721 on Build GRCh38
rs775993470 Chr.21: 33616882-33616882 on Build GRCh38
rs755252342 Chr.21: 33616694-33616694 on Build GRCh38
rs904403375 Chr.21: 33616707-33616707 on Build GRCh38
rs987566725 Chr.21: 33616872-33616872 on Build GRCh38
rs201888688 Chr.21: 33616788-33616788 on Build GRCh38
rs1000109091 Chr.21: 33616758-33616758 on Build GRCh38

View Full Product Details