Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 80513623-80513872 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM151B
Entrez Gene ID
167555
Gene Name
family with sequence similarity 151 member B
Gene Aliases
UNQ9217
UniGene
Hs.338182
Gene Chromosome Location
Chr.5: 80487981-80542563 on Build GRCh38
Transcripts
NM_205548.2 XM_006714564.3 XM_011543234.2 XM_011543235.2 XM_011543236.2 XM_011543237.2 XM_017009166.1 XM_017009167.1 XM_017009168.1 XM_017009169.1 XM_017009170.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6658649 c.229delC p.P78fs*13 Chr.5: 80513681-80513681 on Build GRCh38
4682616 c.283delA p.S96fs*15 Chr.5: 80513735-80513735 on Build GRCh38
6658646 c.290A>T p.N97I Chr.5: 80513742-80513742 on Build GRCh38
1438878 c.174T>C p.A58A Chr.5: 80513626-80513626 on Build GRCh38
4654008 c.228T>C p.H76H Chr.5: 80513680-80513680 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs889798706 Chr.5: 80513680-80513680 on Build GRCh38
rs142396268 Chr.5: 80513744-80513744 on Build GRCh38
rs879175162 Chr.5: 80513832-80513832 on Build GRCh38
rs773404363 Chr.5: 80513714-80513714 on Build GRCh38
rs749591608 Chr.5: 80513691-80513691 on Build GRCh38
rs249011 Chr.5: 80513839-80513839 on Build GRCh38
rs148781457 Chr.5: 80513739-80513739 on Build GRCh38
rs769873660 Chr.5: 80513790-80513790 on Build GRCh38
rs202246220 Chr.5: 80513769-80513769 on Build GRCh38
rs745381322 Chr.5: 80513756-80513756 on Build GRCh38
rs756620090 Chr.5: 80513759-80513759 on Build GRCh38
rs757991789 Chr.5: 80513650-80513650 on Build GRCh38
rs201855456 Chr.5: 80513747-80513747 on Build GRCh38
rs116061206 Chr.5: 80513705-80513705 on Build GRCh38
rs200002361 Chr.5: 80513662-80513662 on Build GRCh38
rs942930955 Chr.5: 80513674-80513674 on Build GRCh38
rs756746791 Chr.5: 80513666-80513666 on Build GRCh38
rs752554182 Chr.5: 80513734-80513734 on Build GRCh38
rs113601961 Chr.5: 80513658-80513658 on Build GRCh38
rs760529705 Chr.5: 80513801-80513801 on Build GRCh38
rs769227897 Chr.5: 80513800-80513801 on Build GRCh38
rs144031733 Chr.5: 80513807-80513807 on Build GRCh38
rs770275789 Chr.5: 80513675-80513675 on Build GRCh38
rs370464941 Chr.5: 80513784-80513784 on Build GRCh38
rs751294941 Chr.5: 80513814-80513814 on Build GRCh38
rs201910623 Chr.5: 80513830-80513831 on Build GRCh38
rs139629089 Chr.5: 80513649-80513649 on Build GRCh38
rs200155440 Chr.5: 80513659-80513659 on Build GRCh38
rs151333640 Chr.5: 80513746-80513746 on Build GRCh38
rs375932934 Chr.5: 80513786-80513786 on Build GRCh38
rs769024987 Chr.5: 80513783-80513783 on Build GRCh38
rs918657918 Chr.5: 80513644-80513644 on Build GRCh38
rs959014785 Chr.5: 80513785-80513785 on Build GRCh38
rs775524405 Chr.5: 80513791-80513791 on Build GRCh38
rs76750557 Chr.5: 80513831-80513831 on Build GRCh38
rs528045525 Chr.5: 80513799-80513799 on Build GRCh38
rs759613582 Chr.5: 80513724-80513724 on Build GRCh38
rs749638025 Chr.5: 80513780-80513780 on Build GRCh38
rs995073056 Chr.5: 80513733-80513733 on Build GRCh38
rs780535078 Chr.5: 80513673-80513673 on Build GRCh38
rs144243505 Chr.5: 80513696-80513696 on Build GRCh38
rs761830086 Chr.5: 80513710-80513710 on Build GRCh38
rs963367412 Chr.5: 80513721-80513721 on Build GRCh38
rs199882067 Chr.5: 80513839-80513839 on Build GRCh38
rs760779236 Chr.5: 80513789-80513789 on Build GRCh38
rs370441276 Chr.5: 80513676-80513676 on Build GRCh38

View Full Product Details