Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 57172513-57172738 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAG2
Entrez Gene ID
9532
Gene Name
BCL2 associated athanogene 2
Gene Aliases
BAG-2, dJ417I1.2
UniGene
Hs.745046
Gene Chromosome Location
Chr.6: 57172306-57185215 on Build GRCh38
Transcripts
NM_004282.3 XM_011514998.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3374355 c.33C>G p.N11K Chr.6: 57172730-57172730 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs971044388 Chr.6: 57172545-57172545 on Build GRCh38
rs375792946 Chr.6: 57172650-57172650 on Build GRCh38
rs532760777 Chr.6: 57172599-57172599 on Build GRCh38
rs537460136 Chr.6: 57172541-57172541 on Build GRCh38
rs1063244 Chr.6: 57172618-57172618 on Build GRCh38
rs964825668 Chr.6: 57172533-57172533 on Build GRCh38
rs139346810 Chr.6: 57172709-57172709 on Build GRCh38
rs749287958 Chr.6: 57172661-57172661 on Build GRCh38
rs764194506 Chr.6: 57172686-57172686 on Build GRCh38
rs774044538 Chr.6: 57172679-57172679 on Build GRCh38
rs369085462 Chr.6: 57172715-57172715 on Build GRCh38
rs1027834957 Chr.6: 57172562-57172562 on Build GRCh38
rs758857860 Chr.6: 57172706-57172706 on Build GRCh38
rs979564719 Chr.6: 57172654-57172654 on Build GRCh38
rs746014326 Chr.6: 57172664-57172682 on Build GRCh38
rs867808556 Chr.6: 57172624-57172624 on Build GRCh38
rs1050874519 Chr.6: 57172701-57172701 on Build GRCh38
rs553332306 Chr.6: 57172647-57172647 on Build GRCh38
rs552510876 Chr.6: 57172634-57172634 on Build GRCh38
rs765711654 Chr.6: 57172696-57172696 on Build GRCh38
rs777447219 Chr.6: 57172714-57172714 on Build GRCh38
rs995294966 Chr.6: 57172544-57172544 on Build GRCh38
rs1035300077 Chr.6: 57172625-57172625 on Build GRCh38
rs752438009 Chr.6: 57172708-57172708 on Build GRCh38
rs990540369 Chr.6: 57172631-57172631 on Build GRCh38
rs977046729 Chr.6: 57172569-57172569 on Build GRCh38
rs570861965 Chr.6: 57172594-57172594 on Build GRCh38
rs960940851 Chr.6: 57172627-57172627 on Build GRCh38
rs762342553 Chr.6: 57172691-57172691 on Build GRCh38
rs372582153 Chr.6: 57172684-57172684 on Build GRCh38
rs370222906 Chr.6: 57172683-57172683 on Build GRCh38
rs777608616 Chr.6: 57172716-57172716 on Build GRCh38
rs76119740 Chr.6: 57172690-57172690 on Build GRCh38
rs923317569 Chr.6: 57172648-57172648 on Build GRCh38
rs770943553 Chr.6: 57172675-57172675 on Build GRCh38
rs990417417 Chr.6: 57172606-57172606 on Build GRCh38
rs558966479 Chr.6: 57172555-57172555 on Build GRCh38
rs1031588304 Chr.6: 57172595-57172595 on Build GRCh38
rs750768984 Chr.6: 57172705-57172705 on Build GRCh38
rs760878662 Chr.6: 57172685-57172685 on Build GRCh38

View Full Product Details