Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 171468843-171469102 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCAF17
Entrez Gene ID
80067
Gene Name
DDB1 and CUL4 associated factor 17
Gene Aliases
C2orf37
UniGene
Hs.659439
Gene Chromosome Location
Chr.2: 171434166-171491029 on Build GRCh38
Transcripts
NM_001164821.1 NM_025000.3 NR_028482.1 XM_006712767.1 XM_006712768.1 XM_011511881.1 XM_011511882.1 XM_017004995.1 XM_017004996.1 XM_017004997.1 XM_017004998.1 XM_017004999.1 XM_017005000.1 XM_017005001.1 XM_017005002.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7336380 c.951C>A p.F317L Chr.2: 171469000-171469000 on Build GRCh38
1691389 c.952C>T p.H318Y Chr.2: 171469001-171469001 on Build GRCh38
5619211 c.939G>A p.Q313Q Chr.2: 171468988-171468988 on Build GRCh38
7116130 c.859G>C p.E287Q Chr.2: 171468908-171468908 on Build GRCh38
3837412 c.961G>A p.A321T Chr.2: 171469010-171469010 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747944118 Chr.2: 171468911-171468911 on Build GRCh38
rs765172251 Chr.2: 171469049-171469050 on Build GRCh38
rs201630526 Chr.2: 171469063-171469063 on Build GRCh38
rs373435062 Chr.2: 171468890-171468890 on Build GRCh38
rs781056426 Chr.2: 171469040-171469040 on Build GRCh38
rs776939642 Chr.2: 171469068-171469068 on Build GRCh38
rs199761860 Chr.2: 171468879-171468879 on Build GRCh38
rs775685832 Chr.2: 171469062-171469062 on Build GRCh38
rs61731491 Chr.2: 171468988-171468988 on Build GRCh38
rs771178201 Chr.2: 171468913-171468913 on Build GRCh38
rs114419034 Chr.2: 171468962-171468962 on Build GRCh38
rs370048164 Chr.2: 171468937-171468937 on Build GRCh38
rs552769867 Chr.2: 171468904-171468904 on Build GRCh38
rs930587020 Chr.2: 171469013-171469013 on Build GRCh38
rs150187836 Chr.2: 171468965-171468965 on Build GRCh38
rs925342589 Chr.2: 171468870-171468870 on Build GRCh38
rs751111716 Chr.2: 171469024-171469024 on Build GRCh38
rs368493049 Chr.2: 171469039-171469039 on Build GRCh38
rs760680285 Chr.2: 171469040-171469040 on Build GRCh38
rs765837085 Chr.2: 171468999-171468999 on Build GRCh38
rs749990174 Chr.2: 171468976-171468976 on Build GRCh38
rs748120696 Chr.2: 171469042-171469042 on Build GRCh38
rs750345667 Chr.2: 171469053-171469054 on Build GRCh38
rs761229686 Chr.2: 171468955-171468955 on Build GRCh38
rs777645107 Chr.2: 171469051-171469051 on Build GRCh38
rs746101867 Chr.2: 171468919-171468919 on Build GRCh38
rs779556352 Chr.2: 171468902-171468902 on Build GRCh38
rs575563191 Chr.2: 171469054-171469054 on Build GRCh38
rs146745875 Chr.2: 171468964-171468964 on Build GRCh38
rs781010136 Chr.2: 171468910-171468910 on Build GRCh38
rs746899310 Chr.2: 171468903-171468903 on Build GRCh38
rs772459391 Chr.2: 171469059-171469059 on Build GRCh38
rs757839826 Chr.2: 171468893-171468893 on Build GRCh38
rs375662120 Chr.2: 171468918-171468918 on Build GRCh38
rs764555678 Chr.2: 171468961-171468961 on Build GRCh38
rs755806887 Chr.2: 171469044-171469044 on Build GRCh38
rs772225477 Chr.2: 171468922-171468922 on Build GRCh38
rs919887099 Chr.2: 171469011-171469011 on Build GRCh38
rs769204619 Chr.2: 171469065-171469065 on Build GRCh38

View Full Product Details