Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 44232276-44232549 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFHC2
Entrez Gene ID
80258
Gene Name
EF-hand domain containing 2
Gene Aliases
MRX74, dJ1158H2.1
UniGene
Hs.521953
Gene Chromosome Location
Chr.X: 44146254-44343677 on Build GRCh38
Transcripts
NM_025184.3 XM_006724562.2

SNP Detail

SNP ID Chromosome Location
rs985628040 Chr.X: 44232305-44232306 on Build GRCh38
rs748673890 Chr.X: 44232438-44232438 on Build GRCh38
rs187962473 Chr.X: 44232406-44232406 on Build GRCh38
rs776513079 Chr.X: 44232393-44232393 on Build GRCh38
rs941778642 Chr.X: 44232371-44232371 on Build GRCh38
rs771239998 Chr.X: 44232491-44232491 on Build GRCh38
rs774755259 Chr.X: 44232498-44232498 on Build GRCh38
rs891696446 Chr.X: 44232451-44232451 on Build GRCh38
rs193126389 Chr.X: 44232436-44232436 on Build GRCh38
rs545372479 Chr.X: 44232339-44232339 on Build GRCh38
rs777944095 Chr.X: 44232465-44232465 on Build GRCh38
rs763399150 Chr.X: 44232486-44232486 on Build GRCh38
rs150556905 Chr.X: 44232303-44232303 on Build GRCh38
rs185264618 Chr.X: 44232437-44232437 on Build GRCh38
rs965614126 Chr.X: 44232317-44232317 on Build GRCh38
rs911410791 Chr.X: 44232316-44232316 on Build GRCh38
rs983098753 Chr.X: 44232308-44232308 on Build GRCh38
rs908876595 Chr.X: 44232335-44232335 on Build GRCh38
rs918847278 Chr.X: 44232345-44232345 on Build GRCh38
rs748361696 Chr.X: 44232449-44232449 on Build GRCh38

View Full Product Details