Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 119520026-119520291 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC60
Entrez Gene ID
160777
Gene Name
coiled-coil domain containing 60
Gene Aliases
-
UniGene
Hs.98188
Gene Chromosome Location
Chr.12: 119334712-119541047 on Build GRCh38
Transcripts
NM_178499.4 XM_017018911.1 XM_017018912.1 XM_017018913.1 XM_017018914.1 XM_017018915.1 XM_017018916.1

Gene Symbol
LOC105370027
Entrez Gene ID
105370027
Gene Name
uncharacterized LOC105370027
Gene Aliases
-
UniGene
Hs.720596
Gene Chromosome Location
Chr.12: 119387991-119668006 on Build GRCh38
Transcripts
NM_178499.4 XM_017018911.1 XM_017018912.1 XM_017018913.1 XM_017018914.1 XM_017018915.1 XM_017018916.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4563616 c.981G>A p.V327V Chr.12: 119520133-119520133 on Build GRCh38
3703887 c.1040+9G>C p.? Chr.12: 119520201-119520201 on Build GRCh38
7093100 c.1004C>A p.S335Y Chr.12: 119520156-119520156 on Build GRCh38
5374339 c.1023G>A p.Q341Q Chr.12: 119520175-119520175 on Build GRCh38
3456895 c.1015G>A p.E339K Chr.12: 119520167-119520167 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754272405 Chr.12: 119520240-119520240 on Build GRCh38
rs764987237 Chr.12: 119520206-119520206 on Build GRCh38
rs116153736 Chr.12: 119520170-119520170 on Build GRCh38
rs746011054 Chr.12: 119520145-119520145 on Build GRCh38
rs764587295 Chr.12: 119520239-119520239 on Build GRCh38
rs368870463 Chr.12: 119520227-119520227 on Build GRCh38
rs756555141 Chr.12: 119520108-119520108 on Build GRCh38
rs708884 Chr.12: 119520056-119520056 on Build GRCh38
rs897150680 Chr.12: 119520261-119520261 on Build GRCh38
rs765854055 Chr.12: 119520146-119520147 on Build GRCh38
rs115862562 Chr.12: 119520090-119520090 on Build GRCh38
rs778828974 Chr.12: 119520138-119520138 on Build GRCh38
rs768065980 Chr.12: 119520226-119520226 on Build GRCh38
rs984541999 Chr.12: 119520148-119520148 on Build GRCh38
rs750462883 Chr.12: 119520149-119520149 on Build GRCh38
rs772580994 Chr.12: 119520218-119520218 on Build GRCh38
rs757653567 Chr.12: 119520080-119520080 on Build GRCh38
rs762361806 Chr.12: 119520137-119520137 on Build GRCh38
rs771984543 Chr.12: 119520147-119520147 on Build GRCh38
rs771006382 Chr.12: 119520133-119520133 on Build GRCh38
rs761339452 Chr.12: 119520182-119520182 on Build GRCh38
rs768461791 Chr.12: 119520171-119520171 on Build GRCh38
rs527882060 Chr.12: 119520054-119520054 on Build GRCh38
rs775473720 Chr.12: 119520157-119520157 on Build GRCh38
rs375108096 Chr.12: 119520114-119520114 on Build GRCh38
rs762587188 Chr.12: 119520223-119520223 on Build GRCh38
rs565882967 Chr.12: 119520250-119520250 on Build GRCh38
rs946698814 Chr.12: 119520265-119520265 on Build GRCh38
rs376471070 Chr.12: 119520089-119520089 on Build GRCh38
rs150119909 Chr.12: 119520107-119520107 on Build GRCh38
rs1023262688 Chr.12: 119520131-119520131 on Build GRCh38
rs889825436 Chr.12: 119520264-119520264 on Build GRCh38
rs755414516 Chr.12: 119520091-119520091 on Build GRCh38
rs778087938 Chr.12: 119520111-119520111 on Build GRCh38
rs776304268 Chr.12: 119520181-119520181 on Build GRCh38
rs573590 Chr.12: 119520263-119520263 on Build GRCh38
rs561585954 Chr.12: 119520060-119520060 on Build GRCh38
rs756501980 Chr.12: 119520232-119520232 on Build GRCh38
rs781778508 Chr.12: 119520103-119520103 on Build GRCh38
rs934239199 Chr.12: 119520230-119520230 on Build GRCh38
rs530330323 Chr.12: 119520069-119520069 on Build GRCh38

View Full Product Details