Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 33014237-33014492 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLB1
Entrez Gene ID
2720
Gene Name
galactosidase beta 1
Gene Aliases
EBP, ELNR1, MPS4B
UniGene
Hs.443031
Gene Chromosome Location
Chr.3: 32996608-33097230 on Build GRCh38
Transcripts
NM_000404.3 NM_001079811.2 NM_001135602.2 NM_001317040.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3767400 c.1528A>T p.T510S Chr.3: 33014262-33014262 on Build GRCh38
302303 c.1530G>A p.T510T Chr.3: 33014260-33014260 on Build GRCh38
4870664 c.1674G>A p.T558T Chr.3: 33014260-33014260 on Build GRCh38
4818446 c.1527G>A p.W509* Chr.3: 33014263-33014263 on Build GRCh38
4818669 c.1543G>C p.D515H Chr.3: 33014247-33014247 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs73826340 Chr.3: 33014384-33014384 on Build GRCh38
rs748176346 Chr.3: 33014272-33014272 on Build GRCh38
rs893047165 Chr.3: 33014416-33014416 on Build GRCh38
rs374924493 Chr.3: 33014269-33014269 on Build GRCh38
rs773182845 Chr.3: 33014286-33014286 on Build GRCh38
rs754228928 Chr.3: 33014357-33014357 on Build GRCh38
rs57857629 Chr.3: 33014459-33014459 on Build GRCh38
rs1002557344 Chr.3: 33014450-33014450 on Build GRCh38
rs1054351579 Chr.3: 33014403-33014403 on Build GRCh38
rs757882979 Chr.3: 33014318-33014318 on Build GRCh38
rs1041770920 Chr.3: 33014441-33014441 on Build GRCh38
rs762655673 Chr.3: 33014289-33014289 on Build GRCh38
rs755911393 Chr.3: 33014392-33014392 on Build GRCh38
rs202007119 Chr.3: 33014309-33014309 on Build GRCh38
rs372538696 Chr.3: 33014279-33014279 on Build GRCh38
rs745555723 Chr.3: 33014402-33014402 on Build GRCh38
rs72555363 Chr.3: 33014263-33014263 on Build GRCh38
rs369598326 Chr.3: 33014270-33014270 on Build GRCh38
rs1803198 Chr.3: 33014261-33014261 on Build GRCh38
rs983190562 Chr.3: 33014340-33014340 on Build GRCh38
rs757418009 Chr.3: 33014358-33014358 on Build GRCh38
rs587776526 Chr.3: 33014312-33014312 on Build GRCh38
rs921750168 Chr.3: 33014397-33014397 on Build GRCh38
rs999417429 Chr.3: 33014454-33014454 on Build GRCh38
rs906909237 Chr.3: 33014446-33014446 on Build GRCh38
rs13073546 Chr.3: 33014259-33014259 on Build GRCh38
rs981899210 Chr.3: 33014278-33014278 on Build GRCh38
rs1034088583 Chr.3: 33014451-33014451 on Build GRCh38
rs541229318 Chr.3: 33014368-33014368 on Build GRCh38
rs764408792 Chr.3: 33014353-33014353 on Build GRCh38
rs753129246 Chr.3: 33014318-33014318 on Build GRCh38
rs929129635 Chr.3: 33014315-33014315 on Build GRCh38
rs933103250 Chr.3: 33014434-33014434 on Build GRCh38
rs191203702 Chr.3: 33014260-33014260 on Build GRCh38
rs761217705 Chr.3: 33014342-33014342 on Build GRCh38
rs943266125 Chr.3: 33014379-33014379 on Build GRCh38
rs1009872931 Chr.3: 33014435-33014435 on Build GRCh38
rs377063117 Chr.3: 33014314-33014314 on Build GRCh38
rs72555368 Chr.3: 33014292-33014292 on Build GRCh38

View Full Product Details