Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 67402685-67402930 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ETAA1
Entrez Gene ID
54465
Gene Name
Ewing tumor associated antigen 1
Gene Aliases
ETAA16
UniGene
Hs.353022
Gene Chromosome Location
Chr.2: 67397310-67410411 on Build GRCh38
Transcripts
NM_019002.3 XM_017004376.1 XM_017004377.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
354311 c.491G>T p.C164F Chr.2: 67402923-67402923 on Build GRCh38
325276 c.476A>G p.E159G Chr.2: 67402908-67402908 on Build GRCh38
5934519 c.430G>A p.D144N Chr.2: 67402862-67402862 on Build GRCh38
6092145 c.488C>T p.P163L Chr.2: 67402920-67402920 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs771249312 Chr.2: 67402872-67402874 on Build GRCh38
rs887742084 Chr.2: 67402809-67402809 on Build GRCh38
rs1003310237 Chr.2: 67402858-67402858 on Build GRCh38
rs774906210 Chr.2: 67402724-67402724 on Build GRCh38
rs1023486836 Chr.2: 67402823-67402823 on Build GRCh38
rs994097132 Chr.2: 67402750-67402750 on Build GRCh38
rs553175123 Chr.2: 67402765-67402765 on Build GRCh38
rs546467595 Chr.2: 67402793-67402793 on Build GRCh38
rs944828142 Chr.2: 67402727-67402727 on Build GRCh38
rs577501655 Chr.2: 67402790-67402790 on Build GRCh38
rs923184 Chr.2: 67402826-67402826 on Build GRCh38
rs199922148 Chr.2: 67402862-67402862 on Build GRCh38
rs541435601 Chr.2: 67402740-67402740 on Build GRCh38
rs373692063 Chr.2: 67402901-67402901 on Build GRCh38
rs1016976903 Chr.2: 67402716-67402716 on Build GRCh38
rs573076431 Chr.2: 67402810-67402810 on Build GRCh38
rs747627324 Chr.2: 67402904-67402904 on Build GRCh38
rs114885838 Chr.2: 67402895-67402895 on Build GRCh38
rs1049053056 Chr.2: 67402795-67402796 on Build GRCh38
rs778418458 Chr.2: 67402903-67402903 on Build GRCh38
rs370615336 Chr.2: 67402846-67402846 on Build GRCh38
rs754386482 Chr.2: 67402867-67402867 on Build GRCh38
rs988261030 Chr.2: 67402902-67402902 on Build GRCh38
rs201042009 Chr.2: 67402886-67402886 on Build GRCh38
rs761052430 Chr.2: 67402851-67402851 on Build GRCh38
rs766836207 Chr.2: 67402866-67402866 on Build GRCh38
rs761590504 Chr.2: 67402821-67402821 on Build GRCh38
rs780117575 Chr.2: 67402893-67402893 on Build GRCh38
rs765416302 Chr.2: 67402876-67402876 on Build GRCh38
rs758581910 Chr.2: 67402878-67402878 on Build GRCh38
rs897892231 Chr.2: 67402787-67402788 on Build GRCh38
rs762648744 Chr.2: 67402820-67402820 on Build GRCh38
rs752859421 Chr.2: 67402877-67402877 on Build GRCh38
rs759512621 Chr.2: 67402871-67402871 on Build GRCh38
rs778161546 Chr.2: 67402880-67402880 on Build GRCh38

View Full Product Details