Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1244158-1244425 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP5D
Entrez Gene ID
513
Gene Name
ATP synthase, H+ transporting, mitochondrial F1 complex, delta subunit
Gene Aliases
-
UniGene
Hs.418668
Gene Chromosome Location
Chr.19: 1241750-1244825 on Build GRCh38
Transcripts
NM_001001975.1 NM_001687.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7315703 c.439G>C p.E147Q Chr.19: 1244369-1244369 on Build GRCh38
6049475 c.452C>T p.A151V Chr.19: 1244382-1244382 on Build GRCh38
1611578 c.381G>A p.L127L Chr.19: 1244182-1244182 on Build GRCh38
5801369 c.416_417ins22 p.E140fs*7 Chr.19: 1244346-1244347 on Build GRCh38
4780622 c.439G>T p.E147* Chr.19: 1244369-1244369 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749690286 Chr.19: 1244301-1244301 on Build GRCh38
rs747636949 Chr.19: 1244384-1244384 on Build GRCh38
rs763552056 Chr.19: 1244206-1244206 on Build GRCh38
rs928011879 Chr.19: 1244376-1244376 on Build GRCh38
rs978153054 Chr.19: 1244375-1244375 on Build GRCh38
rs369774253 Chr.19: 1244353-1244353 on Build GRCh38
rs200485963 Chr.19: 1244382-1244382 on Build GRCh38
rs754880094 Chr.19: 1244342-1244342 on Build GRCh38
rs778239491 Chr.19: 1244300-1244300 on Build GRCh38
rs750127098 Chr.19: 1244336-1244336 on Build GRCh38
rs1018757225 Chr.19: 1244335-1244335 on Build GRCh38
rs750251528 Chr.19: 1244267-1244267 on Build GRCh38
rs780021515 Chr.19: 1244270-1244270 on Build GRCh38
rs1003385367 Chr.19: 1244333-1244334 on Build GRCh38
rs765380938 Chr.19: 1244390-1244390 on Build GRCh38
rs966297306 Chr.19: 1244370-1244370 on Build GRCh38
rs373882473 Chr.19: 1244368-1244368 on Build GRCh38
rs761415689 Chr.19: 1244330-1244330 on Build GRCh38
rs748451782 Chr.19: 1244179-1244179 on Build GRCh38
rs200593395 Chr.19: 1244402-1244402 on Build GRCh38
rs760208758 Chr.19: 1244204-1244204 on Build GRCh38
rs76660329 Chr.19: 1244201-1244201 on Build GRCh38
rs776123691 Chr.19: 1244323-1244323 on Build GRCh38
rs372935723 Chr.19: 1244404-1244404 on Build GRCh38
rs771247266 Chr.19: 1244309-1244309 on Build GRCh38
rs371298161 Chr.19: 1244310-1244310 on Build GRCh38
rs773721236 Chr.19: 1244193-1244193 on Build GRCh38
rs751504967 Chr.19: 1244340-1244340 on Build GRCh38
rs1019507942 Chr.19: 1244369-1244371 on Build GRCh38
rs202216827 Chr.19: 1244268-1244268 on Build GRCh38
rs368262732 Chr.19: 1244237-1244237 on Build GRCh38
rs376783286 Chr.19: 1244333-1244333 on Build GRCh38
rs4476280 Chr.19: 1244280-1244280 on Build GRCh38
rs762781339 Chr.19: 1244338-1244338 on Build GRCh38
rs762573715 Chr.19: 1244397-1244397 on Build GRCh38
rs543979394 Chr.19: 1244250-1244250 on Build GRCh38
rs961669963 Chr.19: 1244351-1244351 on Build GRCh38
rs201047760 Chr.19: 1244276-1244276 on Build GRCh38
rs770265257 Chr.19: 1244188-1244188 on Build GRCh38
rs569682239 Chr.19: 1244366-1244366 on Build GRCh38
rs770142305 Chr.19: 1244299-1244299 on Build GRCh38
rs769336953 Chr.19: 1244396-1244396 on Build GRCh38
rs372215170 Chr.19: 1244265-1244265 on Build GRCh38
rs766957056 Chr.19: 1244306-1244306 on Build GRCh38
rs376321377 Chr.19: 1244377-1244377 on Build GRCh38
rs1055403220 Chr.19: 1244249-1244249 on Build GRCh38
rs144381807 Chr.19: 1244362-1244362 on Build GRCh38
rs747887574 Chr.19: 1244186-1244189 on Build GRCh38
rs912154889 Chr.19: 1244238-1244238 on Build GRCh38
rs1012076152 Chr.19: 1244298-1244301 on Build GRCh38
rs201886813 Chr.19: 1244378-1244378 on Build GRCh38
rs113244457 Chr.19: 1244313-1244313 on Build GRCh38
rs200588209 Chr.19: 1244296-1244296 on Build GRCh38
rs376786572 Chr.19: 1244278-1244278 on Build GRCh38
rs768199132 Chr.19: 1244318-1244318 on Build GRCh38
rs766143982 Chr.19: 1244399-1244399 on Build GRCh38
rs781350269 Chr.19: 1244346-1244346 on Build GRCh38
rs866116135 Chr.19: 1244381-1244381 on Build GRCh38
rs772599030 Chr.19: 1244379-1244379 on Build GRCh38
rs989928611 Chr.19: 1244364-1244364 on Build GRCh38
rs368955968 Chr.19: 1244279-1244279 on Build GRCh38
rs774017721 Chr.19: 1244401-1244401 on Build GRCh38
rs944914625 Chr.19: 1244244-1244244 on Build GRCh38
rs766157566 Chr.19: 1244339-1244339 on Build GRCh38

View Full Product Details