Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 95445904-95446151 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYNE3
Entrez Gene ID
161176
Gene Name
spectrin repeat containing nuclear envelope family member 3
Gene Aliases
C14orf49, NET53, Nesp3
UniGene
Hs.354645
Gene Chromosome Location
Chr.14: 95416083-95516693 on Build GRCh38
Transcripts
NM_152592.3 XM_005267377.3 XM_006720063.3 XM_011536515.2 XM_017021056.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1749053 c.1622G>A p.S541N Chr.14: 95445919-95445919 on Build GRCh38
2253941 c.1594C>T p.L532L Chr.14: 95445947-95445947 on Build GRCh38
2253950 c.1472G>A p.R491H Chr.14: 95446069-95446069 on Build GRCh38
3720381 c.1498C>T p.Q500* Chr.14: 95446043-95446043 on Build GRCh38
4851138 c.1537G>C p.E513Q Chr.14: 95446004-95446004 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759005888 Chr.14: 95445933-95445933 on Build GRCh38
rs757771903 Chr.14: 95446021-95446021 on Build GRCh38
rs377458483 Chr.14: 95446126-95446126 on Build GRCh38
rs776323574 Chr.14: 95446067-95446067 on Build GRCh38
rs752083442 Chr.14: 95445947-95445947 on Build GRCh38
rs572397212 Chr.14: 95445991-95445991 on Build GRCh38
rs778376216 Chr.14: 95445944-95445944 on Build GRCh38
rs778869997 Chr.14: 95446124-95446124 on Build GRCh38
rs147178428 Chr.14: 95445985-95445985 on Build GRCh38
rs373775092 Chr.14: 95446094-95446094 on Build GRCh38
rs755090703 Chr.14: 95446049-95446049 on Build GRCh38
rs772015289 Chr.14: 95445978-95445978 on Build GRCh38
rs759117563 Chr.14: 95445993-95445993 on Build GRCh38
rs764945000 Chr.14: 95445994-95445994 on Build GRCh38
rs574563186 Chr.14: 95446090-95446090 on Build GRCh38
rs780197275 Chr.14: 95446038-95446038 on Build GRCh38
rs140376969 Chr.14: 95446017-95446017 on Build GRCh38
rs756515642 Chr.14: 95446106-95446106 on Build GRCh38
rs576145403 Chr.14: 95446046-95446046 on Build GRCh38
rs770697351 Chr.14: 95445988-95445988 on Build GRCh38
rs769690758 Chr.14: 95446070-95446070 on Build GRCh38
rs372685550 Chr.14: 95446125-95446125 on Build GRCh38
rs542482480 Chr.14: 95445943-95445943 on Build GRCh38
rs369123745 Chr.14: 95446112-95446112 on Build GRCh38
rs779045885 Chr.14: 95446050-95446050 on Build GRCh38
rs150102449 Chr.14: 95446000-95446000 on Build GRCh38
rs767790091 Chr.14: 95446027-95446027 on Build GRCh38
rs17092283 Chr.14: 95446089-95446089 on Build GRCh38
rs990676732 Chr.14: 95445950-95445950 on Build GRCh38
rs749322447 Chr.14: 95445975-95445975 on Build GRCh38
rs535784290 Chr.14: 95445949-95445949 on Build GRCh38
rs766699084 Chr.14: 95446108-95446108 on Build GRCh38
rs200035153 Chr.14: 95446101-95446101 on Build GRCh38
rs1006847434 Chr.14: 95446034-95446034 on Build GRCh38
rs61749972 Chr.14: 95446051-95446051 on Build GRCh38
rs770002936 Chr.14: 95445959-95445959 on Build GRCh38
rs750027767 Chr.14: 95445931-95445931 on Build GRCh38
rs763758960 Chr.14: 95446086-95446086 on Build GRCh38
rs28526182 Chr.14: 95445948-95445948 on Build GRCh38
rs780109883 Chr.14: 95445970-95445970 on Build GRCh38
rs781239512 Chr.14: 95446128-95446128 on Build GRCh38
rs762670639 Chr.14: 95446029-95446030 on Build GRCh38
rs756356344 Chr.14: 95446031-95446031 on Build GRCh38
rs745629148 Chr.14: 95446069-95446069 on Build GRCh38
rs773092090 Chr.14: 95446027-95446027 on Build GRCh38
rs746137627 Chr.14: 95445953-95445953 on Build GRCh38
rs746933327 Chr.14: 95445987-95445987 on Build GRCh38
rs762234500 Chr.14: 95446091-95446091 on Build GRCh38
rs369046578 Chr.14: 95446005-95446005 on Build GRCh38
rs199883379 Chr.14: 95446028-95446028 on Build GRCh38
rs907283226 Chr.14: 95445998-95445998 on Build GRCh38
rs202124505 Chr.14: 95446105-95446105 on Build GRCh38
rs763671081 Chr.14: 95446012-95446012 on Build GRCh38
rs560371407 Chr.14: 95446111-95446111 on Build GRCh38
rs921795608 Chr.14: 95445989-95445989 on Build GRCh38
rs147345417 Chr.14: 95446077-95446077 on Build GRCh38
rs770841219 Chr.14: 95446058-95446058 on Build GRCh38
rs762694667 Chr.14: 95446083-95446083 on Build GRCh38

View Full Product Details