Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155251748-155251995 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM189B
Entrez Gene ID
10712
Gene Name
family with sequence similarity 189 member B
Gene Aliases
C1orf2, COTE1
UniGene
Hs.348308
Gene Chromosome Location
Chr.1: 155247205-155255483 on Build GRCh38
Transcripts
NM_001267608.1 NM_006589.2 NM_198264.1 XM_005244845.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2210296 c.770G>A p.C257Y Chr.1: 155251757-155251757 on Build GRCh38
6229271 c.634C>T p.R212W Chr.1: 155251893-155251893 on Build GRCh38
5730050 c.682C>T p.P228S Chr.1: 155251845-155251845 on Build GRCh38
7194701 c.625G>A p.A209T Chr.1: 155251902-155251902 on Build GRCh38
3376746 c.767C>T p.T256I Chr.1: 155251760-155251760 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs981984128 Chr.1: 155251923-155251923 on Build GRCh38
rs770155126 Chr.1: 155251841-155251841 on Build GRCh38
rs755193895 Chr.1: 155251799-155251799 on Build GRCh38
rs886269225 Chr.1: 155251815-155251815 on Build GRCh38
rs534103219 Chr.1: 155251893-155251893 on Build GRCh38
rs755106714 Chr.1: 155251922-155251922 on Build GRCh38
rs569801826 Chr.1: 155251864-155251864 on Build GRCh38
rs558746279 Chr.1: 155251918-155251918 on Build GRCh38
rs369928505 Chr.1: 155251784-155251784 on Build GRCh38
rs775224298 Chr.1: 155251874-155251874 on Build GRCh38
rs1011323459 Chr.1: 155251908-155251908 on Build GRCh38
rs374300931 Chr.1: 155251856-155251856 on Build GRCh38
rs780055069 Chr.1: 155251791-155251791 on Build GRCh38
rs868415553 Chr.1: 155251858-155251858 on Build GRCh38
rs760481151 Chr.1: 155251875-155251875 on Build GRCh38
rs959193825 Chr.1: 155251954-155251954 on Build GRCh38
rs758548070 Chr.1: 155251789-155251789 on Build GRCh38
rs751552362 Chr.1: 155251919-155251919 on Build GRCh38
rs752397635 Chr.1: 155251772-155251772 on Build GRCh38
rs764049650 Chr.1: 155251892-155251892 on Build GRCh38
rs928836718 Chr.1: 155251925-155251925 on Build GRCh38
rs747232240 Chr.1: 155251798-155251798 on Build GRCh38
rs781506251 Chr.1: 155251806-155251806 on Build GRCh38
rs865898681 Chr.1: 155251862-155251862 on Build GRCh38
rs974931088 Chr.1: 155251944-155251944 on Build GRCh38
rs773778486 Chr.1: 155251853-155251853 on Build GRCh38
rs199852559 Chr.1: 155251850-155251850 on Build GRCh38
rs778029675 Chr.1: 155251849-155251849 on Build GRCh38
rs753832815 Chr.1: 155251788-155251788 on Build GRCh38
rs1033763955 Chr.1: 155251794-155251794 on Build GRCh38
rs895314762 Chr.1: 155251830-155251830 on Build GRCh38
rs1025668417 Chr.1: 155251871-155251871 on Build GRCh38
rs201135304 Chr.1: 155251822-155251822 on Build GRCh38
rs759121051 Chr.1: 155251855-155251855 on Build GRCh38
rs149524897 Chr.1: 155251783-155251783 on Build GRCh38
rs868260628 Chr.1: 155251896-155251896 on Build GRCh38
rs775749818 Chr.1: 155251917-155251917 on Build GRCh38

View Full Product Details