Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 20439542-20439801 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRYL1
Entrez Gene ID
51084
Gene Name
crystallin lambda 1
Gene Aliases
GDH, HEL30, lambda-CRY
UniGene
Hs.370703
Gene Chromosome Location
Chr.13: 20403667-20525873 on Build GRCh38
Transcripts
NM_015974.2 XM_005266415.3 XM_005266416.4 XM_017020624.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6635910 c.382C>T p.P128S Chr.13: 20439649-20439649 on Build GRCh38
6635907 c.280T>C p.C94R Chr.13: 20439751-20439751 on Build GRCh38
243862 c.366C>A p.S122S Chr.13: 20439665-20439665 on Build GRCh38
5023780 c.303_305delGAA p.K104delK Chr.13: 20439726-20439728 on Build GRCh38
4046384 c.405G>T p.L135F Chr.13: 20439626-20439626 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200657659 Chr.13: 20439758-20439758 on Build GRCh38
rs776274999 Chr.13: 20439690-20439690 on Build GRCh38
rs572596378 Chr.13: 20439693-20439693 on Build GRCh38
rs200033152 Chr.13: 20439713-20439713 on Build GRCh38
rs1024100583 Chr.13: 20439579-20439579 on Build GRCh38
rs760274616 Chr.13: 20439687-20439687 on Build GRCh38
rs752983911 Chr.13: 20439615-20439615 on Build GRCh38
rs772956446 Chr.13: 20439685-20439685 on Build GRCh38
rs746158977 Chr.13: 20439668-20439668 on Build GRCh38
rs781239653 Chr.13: 20439652-20439652 on Build GRCh38
rs536324140 Chr.13: 20439762-20439762 on Build GRCh38
rs757760012 Chr.13: 20439768-20439768 on Build GRCh38
rs985232489 Chr.13: 20439624-20439624 on Build GRCh38
rs529923231 Chr.13: 20439631-20439631 on Build GRCh38
rs756817033 Chr.13: 20439718-20439720 on Build GRCh38
rs757273474 Chr.13: 20439645-20439645 on Build GRCh38
rs753137656 Chr.13: 20439717-20439717 on Build GRCh38
rs778103120 Chr.13: 20439621-20439621 on Build GRCh38
rs375891666 Chr.13: 20439604-20439604 on Build GRCh38
rs759313356 Chr.13: 20439691-20439691 on Build GRCh38
rs990922509 Chr.13: 20439572-20439572 on Build GRCh38
rs769874438 Chr.13: 20439670-20439670 on Build GRCh38
rs766132822 Chr.13: 20439595-20439595 on Build GRCh38
rs1006883211 Chr.13: 20439587-20439587 on Build GRCh38
rs765676624 Chr.13: 20439606-20439606 on Build GRCh38
rs753495504 Chr.13: 20439703-20439706 on Build GRCh38
rs763165127 Chr.13: 20439725-20439725 on Build GRCh38
rs907869964 Chr.13: 20439630-20439630 on Build GRCh38
rs764962514 Chr.13: 20439762-20439762 on Build GRCh38
rs367545208 Chr.13: 20439620-20439620 on Build GRCh38
rs747845402 Chr.13: 20439678-20439678 on Build GRCh38
rs139115740 Chr.13: 20439605-20439605 on Build GRCh38
rs772073638 Chr.13: 20439684-20439684 on Build GRCh38
rs778814994 Chr.13: 20439673-20439673 on Build GRCh38
rs183986286 Chr.13: 20439689-20439689 on Build GRCh38
rs922456573 Chr.13: 20439734-20439734 on Build GRCh38
rs199685830 Chr.13: 20439767-20439767 on Build GRCh38
rs1020039343 Chr.13: 20439602-20439602 on Build GRCh38
rs952087515 Chr.13: 20439622-20439622 on Build GRCh38

View Full Product Details