Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65079176-65079427 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDCA5
Entrez Gene ID
113130
Gene Name
cell division cycle associated 5
Gene Aliases
SORORIN
UniGene
Hs.434886
Gene Chromosome Location
Chr.11: 65061009-65084143 on Build GRCh38
Transcripts
NM_080668.3 XM_005273733.3 XM_005273734.3 XM_011544743.2 XM_011544747.2 XM_011544748.2 XM_011544749.2 XM_017017158.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
930310 c.661A>G p.K221E Chr.11: 65079370-65079370 on Build GRCh38
3935572 c.671A>T p.E224V Chr.11: 65079360-65079360 on Build GRCh38
4825840 c.620G>T p.G207V Chr.11: 65079411-65079411 on Build GRCh38
239246 c.685G>A p.E229K Chr.11: 65079181-65079181 on Build GRCh38
415454 c.653A>T p.K218M Chr.11: 65079378-65079378 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757274548 Chr.11: 65079342-65079342 on Build GRCh38
rs746934875 Chr.11: 65079341-65079341 on Build GRCh38
rs753160448 Chr.11: 65079202-65079202 on Build GRCh38
rs994679427 Chr.11: 65079231-65079231 on Build GRCh38
rs149256940 Chr.11: 65079377-65079377 on Build GRCh38
rs781373223 Chr.11: 65079245-65079245 on Build GRCh38
rs771200184 Chr.11: 65079381-65079381 on Build GRCh38
rs774757594 Chr.11: 65079394-65079394 on Build GRCh38
rs1055341605 Chr.11: 65079203-65079203 on Build GRCh38
rs750531620 Chr.11: 65079319-65079319 on Build GRCh38
rs372928302 Chr.11: 65079391-65079391 on Build GRCh38
rs770967585 Chr.11: 65079306-65079306 on Build GRCh38
rs773128401 Chr.11: 65079401-65079401 on Build GRCh38
rs766657504 Chr.11: 65079323-65079323 on Build GRCh38
rs745491639 Chr.11: 65079279-65079279 on Build GRCh38
rs111891963 Chr.11: 65079246-65079246 on Build GRCh38
rs546928568 Chr.11: 65079280-65079281 on Build GRCh38
rs1031650732 Chr.11: 65079371-65079371 on Build GRCh38
rs767483754 Chr.11: 65079312-65079312 on Build GRCh38
rs763391704 Chr.11: 65079213-65079213 on Build GRCh38
rs754071624 Chr.11: 65079381-65079381 on Build GRCh38
rs752495314 Chr.11: 65079334-65079334 on Build GRCh38
rs565801632 Chr.11: 65079278-65079278 on Build GRCh38
rs748979988 Chr.11: 65079382-65079382 on Build GRCh38
rs764535684 Chr.11: 65079301-65079301 on Build GRCh38
rs762375035 Chr.11: 65079307-65079307 on Build GRCh38
rs767880903 Chr.11: 65079325-65079326 on Build GRCh38
rs756227004 Chr.11: 65079322-65079322 on Build GRCh38
rs777878652 Chr.11: 65079274-65079274 on Build GRCh38
rs756418450 Chr.11: 65079250-65079250 on Build GRCh38
rs755178704 Chr.11: 65079403-65079413 on Build GRCh38
rs769499015 Chr.11: 65079291-65079291 on Build GRCh38
rs757306067 Chr.11: 65079230-65079230 on Build GRCh38
rs758249016 Chr.11: 65079335-65079335 on Build GRCh38
rs750850005 Chr.11: 65079334-65079335 on Build GRCh38
rs764587018 Chr.11: 65079219-65079219 on Build GRCh38
rs763338557 Chr.11: 65079298-65079298 on Build GRCh38
rs552327395 Chr.11: 65079272-65079272 on Build GRCh38
rs747963159 Chr.11: 65079271-65079271 on Build GRCh38
rs775971340 Chr.11: 65079345-65079345 on Build GRCh38
rs780398756 Chr.11: 65079369-65079371 on Build GRCh38
rs775648505 Chr.11: 65079297-65079297 on Build GRCh38
rs751598567 Chr.11: 65079226-65079226 on Build GRCh38
rs752243827 Chr.11: 65079204-65079206 on Build GRCh38
rs908186347 Chr.11: 65079217-65079217 on Build GRCh38
rs746988176 Chr.11: 65079276-65079276 on Build GRCh38
rs184338337 Chr.11: 65079403-65079403 on Build GRCh38
rs926076433 Chr.11: 65079317-65079317 on Build GRCh38
rs756457797 Chr.11: 65079344-65079347 on Build GRCh38
rs932518282 Chr.11: 65079282-65079282 on Build GRCh38
rs777747640 Chr.11: 65079336-65079336 on Build GRCh38
rs777095627 Chr.11: 65079395-65079395 on Build GRCh38
rs943818184 Chr.11: 65079225-65079225 on Build GRCh38
rs200209838 Chr.11: 65079364-65079364 on Build GRCh38
rs778518102 Chr.11: 65079265-65079265 on Build GRCh38
rs780506805 Chr.11: 65079348-65079348 on Build GRCh38

View Full Product Details