Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 145868471-145868232 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD35
Entrez Gene ID
148741
Gene Name
ankyrin repeat domain 35
Gene Aliases
-
UniGene
Hs.710624
Gene Chromosome Location
Chr.1: 145866560-145885866 on Build GRCh38
Transcripts
NM_001280799.1 NM_144698.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3360297 c.2862T>C p.L954L Chr.1: 145868326-145868326 on Build GRCh38
7285819 c.2858G>A p.R953H Chr.1: 145868330-145868330 on Build GRCh38
6894465 c.2827G>C p.E943Q Chr.1: 145868361-145868361 on Build GRCh38
674789 c.2874G>T p.L958L Chr.1: 145868314-145868314 on Build GRCh38
5556175 c.2845G>T p.G949* Chr.1: 145868343-145868343 on Build GRCh38
View More

View Full Product Details