Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 117011875-117012130 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD101
Entrez Gene ID
9398
Gene Name
CD101 molecule
Gene Aliases
EWI-101, IGSF2, V7
UniGene
Hs.654598
Gene Chromosome Location
Chr.1: 116990423-117037481 on Build GRCh38
Transcripts
NM_001256106.2 NM_001256109.2 NM_001256111.2 NM_004258.5 XM_017002846.1 XM_017002847.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6045261 c.805C>T p.Q269* Chr.1: 117011930-117011930 on Build GRCh38
4576158 c.756G>A p.T252T Chr.1: 117011881-117011881 on Build GRCh38
6120401 c.810C>T p.T270T Chr.1: 117011935-117011935 on Build GRCh38
4385555 c.824_825insT p.R276fs*16 Chr.1: 117011949-117011950 on Build GRCh38
334174 c.750G>A p.E250E Chr.1: 117011875-117011875 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773163352 Chr.1: 117011935-117011935 on Build GRCh38
rs200728594 Chr.1: 117011898-117011898 on Build GRCh38
rs780364359 Chr.1: 117011897-117011897 on Build GRCh38
rs779350002 Chr.1: 117012077-117012077 on Build GRCh38
rs762472892 Chr.1: 117011970-117011970 on Build GRCh38
rs749560687 Chr.1: 117011900-117011900 on Build GRCh38
rs776557967 Chr.1: 117011944-117011944 on Build GRCh38
rs759333608 Chr.1: 117011955-117011955 on Build GRCh38
rs756593219 Chr.1: 117011992-117011992 on Build GRCh38
rs899602466 Chr.1: 117011903-117011903 on Build GRCh38
rs563556591 Chr.1: 117012080-117012080 on Build GRCh38
rs372656520 Chr.1: 117011936-117011936 on Build GRCh38
rs747890809 Chr.1: 117011924-117011924 on Build GRCh38
rs944992444 Chr.1: 117011948-117011948 on Build GRCh38
rs939136430 Chr.1: 117011934-117011934 on Build GRCh38
rs575637098 Chr.1: 117012100-117012100 on Build GRCh38
rs535176627 Chr.1: 117012031-117012031 on Build GRCh38
rs150217954 Chr.1: 117011922-117011922 on Build GRCh38
rs368209018 Chr.1: 117011999-117011999 on Build GRCh38
rs768852480 Chr.1: 117011902-117011902 on Build GRCh38
rs756459283 Chr.1: 117011896-117011896 on Build GRCh38
rs200178415 Chr.1: 117011982-117011982 on Build GRCh38
rs913583426 Chr.1: 117011947-117011947 on Build GRCh38
rs779030895 Chr.1: 117011917-117011917 on Build GRCh38
rs1044005745 Chr.1: 117012004-117012004 on Build GRCh38
rs958795754 Chr.1: 117012099-117012099 on Build GRCh38
rs149786748 Chr.1: 117012059-117012059 on Build GRCh38
rs866358407 Chr.1: 117011916-117011916 on Build GRCh38
rs764920559 Chr.1: 117011962-117011962 on Build GRCh38
rs771926033 Chr.1: 117011930-117011930 on Build GRCh38
rs754198172 Chr.1: 117012008-117012008 on Build GRCh38
rs1039248966 Chr.1: 117012076-117012076 on Build GRCh38
rs767938011 Chr.1: 117011973-117011973 on Build GRCh38

View Full Product Details