Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 70958178-70958435 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6V1B1
Entrez Gene ID
525
Gene Name
ATPase H+ transporting V1 subunit B1
Gene Aliases
ATP6B1, RTA1B, VATB, VMA2, VPP3
UniGene
Hs.64173
Gene Chromosome Location
Chr.2: 70935868-70965431 on Build GRCh38
Transcripts
NM_001692.3 XM_011532907.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
238903 c.319G>A p.E107K Chr.2: 70958378-70958378 on Build GRCh38
6894570 c.309G>A p.K103K Chr.2: 70958368-70958368 on Build GRCh38
6158615 c.301G>T p.A101S Chr.2: 70958360-70958360 on Build GRCh38
7109822 c.339A>G p.L113L Chr.2: 70958398-70958398 on Build GRCh38
4412076 c.290C>T p.S97L Chr.2: 70958349-70958349 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147933361 Chr.2: 70958356-70958356 on Build GRCh38
rs781825701 Chr.2: 70958414-70958414 on Build GRCh38
rs372140755 Chr.2: 70958320-70958320 on Build GRCh38
rs967063 Chr.2: 70958261-70958261 on Build GRCh38
rs916116968 Chr.2: 70958256-70958256 on Build GRCh38
rs782015931 Chr.2: 70958374-70958374 on Build GRCh38
rs368815802 Chr.2: 70958298-70958298 on Build GRCh38
rs782073814 Chr.2: 70958390-70958390 on Build GRCh38
rs531614845 Chr.2: 70958357-70958357 on Build GRCh38
rs782767344 Chr.2: 70958392-70958392 on Build GRCh38
rs377280267 Chr.2: 70958328-70958328 on Build GRCh38
rs200269431 Chr.2: 70958400-70958400 on Build GRCh38
rs782443411 Chr.2: 70958406-70958406 on Build GRCh38
rs782138777 Chr.2: 70958399-70958399 on Build GRCh38
rs145734697 Chr.2: 70958407-70958407 on Build GRCh38
rs782387976 Chr.2: 70958370-70958370 on Build GRCh38
rs936147818 Chr.2: 70958231-70958231 on Build GRCh38
rs782495957 Chr.2: 70958394-70958394 on Build GRCh38
rs924710147 Chr.2: 70958215-70958215 on Build GRCh38
rs782486724 Chr.2: 70958318-70958318 on Build GRCh38
rs374454025 Chr.2: 70958291-70958291 on Build GRCh38
rs370511629 Chr.2: 70958378-70958378 on Build GRCh38
rs782054296 Chr.2: 70958293-70958293 on Build GRCh38
rs977506073 Chr.2: 70958213-70958213 on Build GRCh38
rs727502893 Chr.2: 70958376-70958376 on Build GRCh38
rs373239487 Chr.2: 70958324-70958324 on Build GRCh38
rs782243410 Chr.2: 70958365-70958365 on Build GRCh38
rs782326782 Chr.2: 70958360-70958360 on Build GRCh38
rs781907249 Chr.2: 70958404-70958404 on Build GRCh38
rs782727362 Chr.2: 70958274-70958274 on Build GRCh38
rs781926766 Chr.2: 70958388-70958388 on Build GRCh38
rs560415314 Chr.2: 70958257-70958257 on Build GRCh38
rs202011016 Chr.2: 70958364-70958364 on Build GRCh38
rs782720902 Chr.2: 70958377-70958377 on Build GRCh38
rs1045991655 Chr.2: 70958279-70958279 on Build GRCh38
rs185226685 Chr.2: 70958249-70958249 on Build GRCh38

View Full Product Details