Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 81965493-81965730 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNA2D1
Entrez Gene ID
781
Gene Name
calcium voltage-gated channel auxiliary subunit alpha2delta 1
Gene Aliases
CACNA2, CACNL2A, CCHL2A, LINC01112, lncRNA-N3
UniGene
Hs.282151
Gene Chromosome Location
Chr.7: 81946444-82443921 on Build GRCh38
Transcripts
NM_000722.3 XM_005250570.2 XM_005250572.2 XM_005250573.2 XM_005250574.2 XM_006716118.2 XM_006716119.3 XM_006716120.3 XM_006716121.3 XM_011516570.2 XM_011516571.2 XM_011516572.2 XM_017012588.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4867170 c.2604T>C p.T868T Chr.7: 81965600-81965600 on Build GRCh38
4783998 c.2597A>T p.D866V Chr.7: 81965607-81965607 on Build GRCh38
5720959 c.2560G>A p.D854N Chr.7: 81965608-81965608 on Build GRCh38
4868131 c.2583G>A p.M861I Chr.7: 81965621-81965621 on Build GRCh38
7249991 c.2584G>T p.A862S Chr.7: 81965620-81965620 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs984635322 Chr.7: 81965538-81965538 on Build GRCh38
rs779525985 Chr.7: 81965705-81965705 on Build GRCh38
rs533025081 Chr.7: 81965591-81965591 on Build GRCh38
rs112134094 Chr.7: 81965639-81965639 on Build GRCh38
rs769258730 Chr.7: 81965546-81965546 on Build GRCh38
rs759559192 Chr.7: 81965682-81965682 on Build GRCh38
rs757283095 Chr.7: 81965545-81965545 on Build GRCh38
rs376388569 Chr.7: 81965691-81965691 on Build GRCh38
rs768922047 Chr.7: 81965558-81965558 on Build GRCh38
rs991824602 Chr.7: 81965549-81965549 on Build GRCh38
rs202062464 Chr.7: 81965544-81965544 on Build GRCh38
rs377103202 Chr.7: 81965675-81965675 on Build GRCh38
rs1015002349 Chr.7: 81965572-81965572 on Build GRCh38
rs767195311 Chr.7: 81965684-81965684 on Build GRCh38
rs78750249 Chr.7: 81965576-81965576 on Build GRCh38
rs775960466 Chr.7: 81965547-81965547 on Build GRCh38
rs760781183 Chr.7: 81965551-81965551 on Build GRCh38
rs751474463 Chr.7: 81965613-81965613 on Build GRCh38
rs774634676 Chr.7: 81965671-81965675 on Build GRCh38
rs996404987 Chr.7: 81965540-81965540 on Build GRCh38
rs756853625 Chr.7: 81965695-81965695 on Build GRCh38
rs373306416 Chr.7: 81965587-81965587 on Build GRCh38
rs755718948 Chr.7: 81965688-81965688 on Build GRCh38

View Full Product Details