Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 20645571-20645832 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCB5
Entrez Gene ID
340273
Gene Name
ATP binding cassette subfamily B member 5
Gene Aliases
ABCB5alpha, ABCB5beta, EST422562
UniGene
Hs.404102
Gene Chromosome Location
Chr.7: 20615622-20757014 on Build GRCh38
Transcripts
NM_001163941.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5413900 c.731C>T p.A244V Chr.7: 20645808-20645808 on Build GRCh38
3832470 c.689C>T p.S230L Chr.7: 20645766-20645766 on Build GRCh38
4968423 c.751T>C p.S251P Chr.7: 20645828-20645828 on Build GRCh38
5026711 c.698G>C p.S233T Chr.7: 20645775-20645775 on Build GRCh38
5931935 c.742G>A p.E248K Chr.7: 20645819-20645819 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776840832 Chr.7: 20645602-20645602 on Build GRCh38
rs112179616 Chr.7: 20645697-20645697 on Build GRCh38
rs775816827 Chr.7: 20645706-20645706 on Build GRCh38
rs902220448 Chr.7: 20645802-20645802 on Build GRCh38
rs1027036638 Chr.7: 20645626-20645626 on Build GRCh38
rs961548292 Chr.7: 20645625-20645625 on Build GRCh38
rs949024215 Chr.7: 20645767-20645767 on Build GRCh38
rs746834535 Chr.7: 20645777-20645777 on Build GRCh38
rs763132245 Chr.7: 20645708-20645708 on Build GRCh38
rs758406026 Chr.7: 20645762-20645762 on Build GRCh38
rs1045158271 Chr.7: 20645698-20645698 on Build GRCh38
rs752113363 Chr.7: 20645757-20645757 on Build GRCh38
rs774285720 Chr.7: 20645714-20645714 on Build GRCh38
rs759432159 Chr.7: 20645718-20645718 on Build GRCh38
rs931218850 Chr.7: 20645729-20645729 on Build GRCh38
rs760223834 Chr.7: 20645617-20645617 on Build GRCh38
rs753429380 Chr.7: 20645734-20645734 on Build GRCh38
rs530497577 Chr.7: 20645719-20645720 on Build GRCh38
rs993855717 Chr.7: 20645625-20645626 on Build GRCh38
rs767880845 Chr.7: 20645717-20645717 on Build GRCh38
rs924225901 Chr.7: 20645662-20645662 on Build GRCh38
rs954689214 Chr.7: 20645684-20645684 on Build GRCh38
rs965575373 Chr.7: 20645651-20645651 on Build GRCh38
rs17216606 Chr.7: 20645628-20645628 on Build GRCh38
rs1032926106 Chr.7: 20645614-20645614 on Build GRCh38
rs910966733 Chr.7: 20645637-20645637 on Build GRCh38
rs770786235 Chr.7: 20645782-20645782 on Build GRCh38
rs989516903 Chr.7: 20645685-20645685 on Build GRCh38
rs985230100 Chr.7: 20645635-20645635 on Build GRCh38
rs77094935 Chr.7: 20645746-20645746 on Build GRCh38
rs754611572 Chr.7: 20645740-20645740 on Build GRCh38
rs762450285 Chr.7: 20645716-20645716 on Build GRCh38
rs760980834 Chr.7: 20645723-20645723 on Build GRCh38
rs916524957 Chr.7: 20645756-20645756 on Build GRCh38
rs151287364 Chr.7: 20645774-20645774 on Build GRCh38
rs560675363 Chr.7: 20645691-20645691 on Build GRCh38
rs558603324 Chr.7: 20645787-20645787 on Build GRCh38
rs976879911 Chr.7: 20645656-20645656 on Build GRCh38
rs527457310 Chr.7: 20645687-20645687 on Build GRCh38
rs766023584 Chr.7: 20645732-20645732 on Build GRCh38
rs756886230 Chr.7: 20645746-20645746 on Build GRCh38
rs1029226492 Chr.7: 20645673-20645673 on Build GRCh38
rs531288225 Chr.7: 20645712-20645712 on Build GRCh38
rs1035608561 Chr.7: 20645609-20645609 on Build GRCh38
rs906621799 Chr.7: 20645713-20645713 on Build GRCh38
rs544948468 Chr.7: 20645621-20645621 on Build GRCh38
rs111890680 Chr.7: 20645690-20645690 on Build GRCh38
rs777882501 Chr.7: 20645715-20645716 on Build GRCh38

View Full Product Details