Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 74045925-74046186 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BBOF1
Entrez Gene ID
80127
Gene Name
basal body orientation factor 1
Gene Aliases
C14orf45, CCDC176
UniGene
Hs.644621
Gene Chromosome Location
Chr.14: 74019350-74078351 on Build GRCh38
Transcripts
NM_025057.2 XM_005268092.3 XM_005268097.3 XM_005268098.4 XM_011537170.2 XM_011537171.2 XM_011537172.1 XM_011537174.2 XM_011537175.2 XM_011537176.2 XM_011537177.2 XM_011537178.2 XM_011537179.2 XM_017021659.1 XM_017021660.1 XM_017021661.1 XM_017021662.1 XM_017021663.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3420025 c.603G>T p.K201N Chr.14: 74046086-74046086 on Build GRCh38
5478752 c.628G>A p.A210T Chr.14: 74046111-74046111 on Build GRCh38
4401827 c.629C>T p.A210V Chr.14: 74046112-74046112 on Build GRCh38
3956209 c.590A>T p.Q197L Chr.14: 74046073-74046073 on Build GRCh38
4666866 c.614T>C p.M205T Chr.14: 74046097-74046097 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761757358 Chr.14: 74046144-74046144 on Build GRCh38
rs748598114 Chr.14: 74046080-74046080 on Build GRCh38
rs751441701 Chr.14: 74046043-74046043 on Build GRCh38
rs937534701 Chr.14: 74045956-74045956 on Build GRCh38
rs183771610 Chr.14: 74046092-74046092 on Build GRCh38
rs950326148 Chr.14: 74046140-74046140 on Build GRCh38
rs983416288 Chr.14: 74046157-74046157 on Build GRCh38
rs745320500 Chr.14: 74046091-74046091 on Build GRCh38
rs750449804 Chr.14: 74046063-74046063 on Build GRCh38
rs772640645 Chr.14: 74046085-74046085 on Build GRCh38
rs764713541 Chr.14: 74046090-74046091 on Build GRCh38
rs376489662 Chr.14: 74046027-74046027 on Build GRCh38
rs550497091 Chr.14: 74046004-74046004 on Build GRCh38
rs1054455643 Chr.14: 74045961-74045961 on Build GRCh38
rs780732509 Chr.14: 74046025-74046025 on Build GRCh38
rs779530578 Chr.14: 74046074-74046074 on Build GRCh38
rs917604171 Chr.14: 74046129-74046129 on Build GRCh38
rs781565684 Chr.14: 74045997-74045998 on Build GRCh38
rs767069719 Chr.14: 74046021-74046021 on Build GRCh38
rs6574158 Chr.14: 74045995-74045995 on Build GRCh38
rs761951448 Chr.14: 74046054-74046054 on Build GRCh38
rs755273535 Chr.14: 74046068-74046068 on Build GRCh38
rs754123762 Chr.14: 74046090-74046092 on Build GRCh38
rs141857440 Chr.14: 74046113-74046113 on Build GRCh38
rs570244556 Chr.14: 74046005-74046005 on Build GRCh38
rs371096556 Chr.14: 74046117-74046117 on Build GRCh38
rs936180883 Chr.14: 74046052-74046052 on Build GRCh38
rs757682256 Chr.14: 74046139-74046145 on Build GRCh38
rs767394238 Chr.14: 74046147-74046147 on Build GRCh38
rs750535152 Chr.14: 74046149-74046149 on Build GRCh38
rs756352111 Chr.14: 74046064-74046064 on Build GRCh38
rs1010188378 Chr.14: 74045980-74045980 on Build GRCh38
rs762866064 Chr.14: 74046115-74046115 on Build GRCh38
rs780859463 Chr.14: 74046086-74046086 on Build GRCh38
rs761042866 Chr.14: 74046084-74046086 on Build GRCh38
rs780100779 Chr.14: 74046066-74046066 on Build GRCh38
rs764129960 Chr.14: 74046042-74046042 on Build GRCh38
rs893255915 Chr.14: 74045967-74045967 on Build GRCh38
rs774132070 Chr.14: 74046130-74046130 on Build GRCh38
rs867000140 Chr.14: 74046095-74046095 on Build GRCh38
rs767592531 Chr.14: 74046060-74046060 on Build GRCh38
rs752283720 Chr.14: 74046022-74046022 on Build GRCh38
rs776492251 Chr.14: 74046016-74046016 on Build GRCh38
rs953228513 Chr.14: 74046065-74046065 on Build GRCh38
rs200660156 Chr.14: 74046041-74046041 on Build GRCh38
rs374031095 Chr.14: 74045996-74045996 on Build GRCh38

View Full Product Details