Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 372461-372726 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B4GALNT4
Entrez Gene ID
338707
Gene Name
beta-1,4-N-acetyl-galactosaminyltransferase 4
Gene Aliases
-
UniGene
Hs.148074
Gene Chromosome Location
Chr.11: 369773-382117 on Build GRCh38
Transcripts
NM_178537.4 XM_017017654.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5838504 c.293G>A p.G98E Chr.11: 372699-372699 on Build GRCh38
466784 c.278T>C p.M93T Chr.11: 372684-372684 on Build GRCh38
3447134 c.259G>A p.E87K Chr.11: 372665-372665 on Build GRCh38
6252983 c.303G>T p.R101S Chr.11: 372709-372709 on Build GRCh38
1353701 c.266G>A p.R89Q Chr.11: 372672-372672 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs992694023 Chr.11: 372604-372604 on Build GRCh38
rs1039483342 Chr.11: 372504-372504 on Build GRCh38
rs545853129 Chr.11: 372650-372650 on Build GRCh38
rs746496907 Chr.11: 372665-372665 on Build GRCh38
rs1023961381 Chr.11: 372605-372605 on Build GRCh38
rs80301163 Chr.11: 372671-372671 on Build GRCh38
rs371925685 Chr.11: 372621-372621 on Build GRCh38
rs530402369 Chr.11: 372695-372695 on Build GRCh38
rs373895499 Chr.11: 372632-372632 on Build GRCh38
rs893822299 Chr.11: 372581-372581 on Build GRCh38
rs942319090 Chr.11: 372499-372499 on Build GRCh38
rs778861629 Chr.11: 372702-372702 on Build GRCh38
rs145123446 Chr.11: 372685-372685 on Build GRCh38
rs182805582 Chr.11: 372701-372701 on Build GRCh38
rs993620902 Chr.11: 372496-372496 on Build GRCh38
rs769881669 Chr.11: 372677-372677 on Build GRCh38
rs761897703 Chr.11: 372618-372618 on Build GRCh38
rs766455442 Chr.11: 372646-372646 on Build GRCh38
rs35475866 Chr.11: 372700-372700 on Build GRCh38
rs766044449 Chr.11: 372486-372486 on Build GRCh38
rs773421318 Chr.11: 372698-372698 on Build GRCh38
rs141645053 Chr.11: 372589-372589 on Build GRCh38
rs1052358372 Chr.11: 372536-372536 on Build GRCh38
rs767831419 Chr.11: 372619-372619 on Build GRCh38
rs755497830 Chr.11: 372653-372653 on Build GRCh38
rs368072591 Chr.11: 372664-372664 on Build GRCh38
rs143485118 Chr.11: 372600-372600 on Build GRCh38
rs756287830 Chr.11: 372645-372645 on Build GRCh38
rs922329527 Chr.11: 372521-372521 on Build GRCh38
rs868747029 Chr.11: 372552-372552 on Build GRCh38
rs540174071 Chr.11: 372543-372543 on Build GRCh38
rs760821783 Chr.11: 372699-372699 on Build GRCh38
rs958641071 Chr.11: 372601-372601 on Build GRCh38
rs562455133 Chr.11: 372659-372659 on Build GRCh38
rs375838832 Chr.11: 372662-372662 on Build GRCh38
rs762867691 Chr.11: 372694-372694 on Build GRCh38
rs11822725 Chr.11: 372669-372669 on Build GRCh38
rs779211038 Chr.11: 372655-372655 on Build GRCh38
rs967018173 Chr.11: 372623-372623 on Build GRCh38
rs1053940363 Chr.11: 372567-372567 on Build GRCh38
rs963733016 Chr.11: 372686-372686 on Build GRCh38
rs758864066 Chr.11: 372707-372707 on Build GRCh38
rs933546454 Chr.11: 372522-372522 on Build GRCh38
rs752560473 Chr.11: 372612-372612 on Build GRCh38
rs191975431 Chr.11: 372672-372672 on Build GRCh38
rs1040426733 Chr.11: 372481-372481 on Build GRCh38

View Full Product Details