Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 124989471-124989744 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTBP2
Entrez Gene ID
1488
Gene Name
C-terminal binding protein 2
Gene Aliases
-
UniGene
Hs.501345
Gene Chromosome Location
Chr.10: 124987849-125161061 on Build GRCh38
Transcripts
NM_001083914.2 NM_001290214.2 NM_001290215.2 NM_001321012.1 NM_001321013.1 NM_001321014.1 NM_001329.3 NM_022802.2 XM_005269561.2 XM_005269564.2 XM_005269567.2 XM_005269568.4 XM_005269569.2 XM_005269571.2 XM_005269572.3 XM_006717642.2 XM_011539349.2 XM_011539351.1 XM_011539353.1 XM_011539354.1 XM_011539355.1 XM_011539357.1 XM_011539358.1 XM_017015756.1 XM_017015757.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4958743 c.2785C>T p.P929S Chr.10: 124989691-124989691 on Build GRCh38
1603115 c.2799T>C p.G933G Chr.10: 124989677-124989677 on Build GRCh38
7310483 c.1338G>A p.*446* Chr.10: 124989518-124989518 on Build GRCh38
4144424 c.1323C>T p.H441H Chr.10: 124989533-124989533 on Build GRCh38
1316963 c.1277C>G p.A426G Chr.10: 124989579-124989579 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745378265 Chr.10: 124989621-124989621 on Build GRCh38
rs762838277 Chr.10: 124989678-124989678 on Build GRCh38
rs146097043 Chr.10: 124989565-124989565 on Build GRCh38
rs73373169 Chr.10: 124989523-124989523 on Build GRCh38
rs145795665 Chr.10: 124989603-124989603 on Build GRCh38
rs779667438 Chr.10: 124989696-124989696 on Build GRCh38
rs767514276 Chr.10: 124989571-124989571 on Build GRCh38
rs773234721 Chr.10: 124989670-124989670 on Build GRCh38
rs755516247 Chr.10: 124989699-124989699 on Build GRCh38
rs78981424 Chr.10: 124989518-124989518 on Build GRCh38
rs75839774 Chr.10: 124989543-124989543 on Build GRCh38
rs77752853 Chr.10: 124989652-124989652 on Build GRCh38
rs35551618 Chr.10: 124989635-124989636 on Build GRCh38
rs753746171 Chr.10: 124989499-124989499 on Build GRCh38
rs113985797 Chr.10: 124989672-124989672 on Build GRCh38
rs151116298 Chr.10: 124989526-124989526 on Build GRCh38
rs140913543 Chr.10: 124989554-124989554 on Build GRCh38
rs796230948 Chr.10: 124989638-124989638 on Build GRCh38
rs202193654 Chr.10: 124989682-124989682 on Build GRCh38
rs759698405 Chr.10: 124989553-124989553 on Build GRCh38
rs748974251 Chr.10: 124989639-124989639 on Build GRCh38
rs369470763 Chr.10: 124989716-124989716 on Build GRCh38
rs756901110 Chr.10: 124989692-124989692 on Build GRCh38
rs201062217 Chr.10: 124989683-124989683 on Build GRCh38
rs369976400 Chr.10: 124989556-124989557 on Build GRCh38
rs767182849 Chr.10: 124989693-124989693 on Build GRCh38
rs756741005 Chr.10: 124989627-124989627 on Build GRCh38
rs141252590 Chr.10: 124989691-124989691 on Build GRCh38
rs929021807 Chr.10: 124989641-124989641 on Build GRCh38
rs758205726 Chr.10: 124989608-124989608 on Build GRCh38
rs1021788055 Chr.10: 124989613-124989613 on Build GRCh38
rs926259434 Chr.10: 124989502-124989502 on Build GRCh38
rs748365238 Chr.10: 124989658-124989658 on Build GRCh38
rs372168977 Chr.10: 124989527-124989527 on Build GRCh38
rs139940617 Chr.10: 124989528-124989528 on Build GRCh38
rs367739342 Chr.10: 124989560-124989560 on Build GRCh38
rs751202687 Chr.10: 124989612-124989612 on Build GRCh38
rs778073304 Chr.10: 124989717-124989717 on Build GRCh38
rs546918459 Chr.10: 124989611-124989611 on Build GRCh38
rs772304305 Chr.10: 124989664-124989664 on Build GRCh38
rs202165277 Chr.10: 124989547-124989547 on Build GRCh38
rs748247495 Chr.10: 124989494-124989495 on Build GRCh38
rs778948717 Chr.10: 124989690-124989692 on Build GRCh38
rs796188515 Chr.10: 124989496-124989496 on Build GRCh38
rs76905908 Chr.10: 124989494-124989494 on Build GRCh38
rs750625993 Chr.10: 124989501-124989501 on Build GRCh38
rs979232406 Chr.10: 124989674-124989674 on Build GRCh38
rs114432763 Chr.10: 124989677-124989677 on Build GRCh38
rs756887482 Chr.10: 124989615-124989615 on Build GRCh38
rs770435602 Chr.10: 124989695-124989696 on Build GRCh38
rs200050704 Chr.10: 124989687-124989687 on Build GRCh38
rs749831405 Chr.10: 124989697-124989697 on Build GRCh38
rs375566697 Chr.10: 124989707-124989707 on Build GRCh38
rs747713301 Chr.10: 124989690-124989691 on Build GRCh38
rs1014894191 Chr.10: 124989551-124989551 on Build GRCh38
rs997739109 Chr.10: 124989567-124989567 on Build GRCh38
rs1048828483 Chr.10: 124989642-124989642 on Build GRCh38
rs142997765 Chr.10: 124989521-124989521 on Build GRCh38
rs780720474 Chr.10: 124989634-124989634 on Build GRCh38
rs146419230 Chr.10: 124989579-124989579 on Build GRCh38
rs373564090 Chr.10: 124989540-124989540 on Build GRCh38
rs746386463 Chr.10: 124989535-124989535 on Build GRCh38
rs780617077 Chr.10: 124989620-124989620 on Build GRCh38
rs796932553 Chr.10: 124989594-124989594 on Build GRCh38
rs75408575 Chr.10: 124989533-124989533 on Build GRCh38
rs766392320 Chr.10: 124989676-124989676 on Build GRCh38
rs765157987 Chr.10: 124989602-124989602 on Build GRCh38
rs771679650 Chr.10: 124989691-124989692 on Build GRCh38
rs749745328 Chr.10: 124989637-124989637 on Build GRCh38
rs76963654 Chr.10: 124989578-124989578 on Build GRCh38
rs755688829 Chr.10: 124989695-124989695 on Build GRCh38
rs775928033 Chr.10: 124989696-124989697 on Build GRCh38
rs768640408 Chr.10: 124989671-124989676 on Build GRCh38
rs368043534 Chr.10: 124989520-124989520 on Build GRCh38
rs76370804 Chr.10: 124989559-124989559 on Build GRCh38
rs748544110 Chr.10: 124989713-124989713 on Build GRCh38
rs769170896 Chr.10: 124989639-124989639 on Build GRCh38
rs909045814 Chr.10: 124989648-124989648 on Build GRCh38
rs749657299 Chr.10: 124989550-124989550 on Build GRCh38
rs578199923 Chr.10: 124989632-124989632 on Build GRCh38
rs78512061 Chr.10: 124989506-124989506 on Build GRCh38
rs796540503 Chr.10: 124989509-124989509 on Build GRCh38
rs762050471 Chr.10: 124989569-124989569 on Build GRCh38

View Full Product Details