Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 158292667-158292902 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD1B
Entrez Gene ID
910
Gene Name
CD1b molecule
Gene Aliases
CD1, CD1A, R1
UniGene
Hs.1310
Gene Chromosome Location
Chr.1: 158286841-158332038 on Build GRCh38
Transcripts
XM_017002785.1 NM_001765.2 XM_005245579.4

Gene Symbol
CD1C
Entrez Gene ID
911
Gene Name
CD1c molecule
Gene Aliases
BDCA1, CD1, CD1A, R7
UniGene
Hs.132448
Gene Chromosome Location
Chr.1: 158289773-158294774 on Build GRCh38
Transcripts
XM_017002785.1 NM_001765.2 XM_005245579.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4559466 c.805G>A p.E269K Chr.1: 158292790-158292790 on Build GRCh38
6459456 c.859C>T p.L287L Chr.1: 158292844-158292844 on Build GRCh38
6339784 c.858T>C p.S286S Chr.1: 158292843-158292843 on Build GRCh38
3976285 c.889+5G>T p.? Chr.1: 158292879-158292879 on Build GRCh38
5796903 c.763C>G p.L255V Chr.1: 158292748-158292748 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758902960 Chr.1: 158292751-158292751 on Build GRCh38
rs765406937 Chr.1: 158292831-158292831 on Build GRCh38
rs762791288 Chr.1: 158292718-158292718 on Build GRCh38
rs989408136 Chr.1: 158292839-158292839 on Build GRCh38
rs150425079 Chr.1: 158292706-158292706 on Build GRCh38
rs757329481 Chr.1: 158292729-158292729 on Build GRCh38
rs149354390 Chr.1: 158292728-158292728 on Build GRCh38
rs775950742 Chr.1: 158292699-158292699 on Build GRCh38
rs766792421 Chr.1: 158292845-158292845 on Build GRCh38
rs146296717 Chr.1: 158292754-158292754 on Build GRCh38
rs773196926 Chr.1: 158292788-158292788 on Build GRCh38
rs770723646 Chr.1: 158292811-158292811 on Build GRCh38
rs766156680 Chr.1: 158292720-158292720 on Build GRCh38
rs3176971 Chr.1: 158292799-158292799 on Build GRCh38
rs746301692 Chr.1: 158292800-158292800 on Build GRCh38
rs780501974 Chr.1: 158292753-158292753 on Build GRCh38
rs867045352 Chr.1: 158292740-158292740 on Build GRCh38
rs750954740 Chr.1: 158292743-158292743 on Build GRCh38
rs201219470 Chr.1: 158292686-158292686 on Build GRCh38
rs376541578 Chr.1: 158292721-158292721 on Build GRCh38
rs780763972 Chr.1: 158292865-158292865 on Build GRCh38
rs139069182 Chr.1: 158292833-158292833 on Build GRCh38
rs768221526 Chr.1: 158292694-158292694 on Build GRCh38
rs3176970 Chr.1: 158292787-158292787 on Build GRCh38
rs144494205 Chr.1: 158292872-158292872 on Build GRCh38
rs373456243 Chr.1: 158292805-158292805 on Build GRCh38
rs969251812 Chr.1: 158292866-158292866 on Build GRCh38
rs866382982 Chr.1: 158292742-158292742 on Build GRCh38
rs41273485 Chr.1: 158292832-158292832 on Build GRCh38
rs772544606 Chr.1: 158292685-158292685 on Build GRCh38
rs760750400 Chr.1: 158292722-158292722 on Build GRCh38
rs375607782 Chr.1: 158292826-158292826 on Build GRCh38
rs754584129 Chr.1: 158292693-158292693 on Build GRCh38
rs147191023 Chr.1: 158292862-158292862 on Build GRCh38
rs982391642 Chr.1: 158292874-158292874 on Build GRCh38
rs111303778 Chr.1: 158292807-158292807 on Build GRCh38
rs115248892 Chr.1: 158292829-158292829 on Build GRCh38
rs770048157 Chr.1: 158292815-158292815 on Build GRCh38
rs3176969 Chr.1: 158292786-158292786 on Build GRCh38
rs139020586 Chr.1: 158292779-158292779 on Build GRCh38
rs751906751 Chr.1: 158292860-158292860 on Build GRCh38
rs776891906 Chr.1: 158292827-158292827 on Build GRCh38
rs1024484957 Chr.1: 158292868-158292868 on Build GRCh38
rs779613937 Chr.1: 158292734-158292734 on Build GRCh38
rs761837821 Chr.1: 158292700-158292700 on Build GRCh38
rs768148459 Chr.1: 158292771-158292771 on Build GRCh38
rs901306034 Chr.1: 158292785-158292785 on Build GRCh38
rs773968611 Chr.1: 158292814-158292814 on Build GRCh38
rs899159182 Chr.1: 158292758-158292758 on Build GRCh38

View Full Product Details