Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31639455-31639700 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAG6
Entrez Gene ID
7917
Gene Name
BCL2 associated athanogene 6
Gene Aliases
BAG-6, BAT3, D6S52E, G3
UniGene
Hs.440900
Gene Chromosome Location
Chr.6: 31639028-31660900 on Build GRCh38
Transcripts
NM_001098534.1 NM_001199697.1 NM_001199698.1 NM_004639.3 NM_080702.2 NM_080703.2 XM_011514892.2 XM_017011275.1 XM_017011276.1 XM_017011277.1 XM_017011278.1 XM_017011279.1 XM_017011280.1 XM_017011281.1 XM_017011282.1 XM_017011283.1 XM_017011284.1 XM_017011285.1 XM_017011286.1 XM_017011287.1 XM_017011288.1 XM_017011289.1 XM_017011290.1 XM_017011291.1 XM_017011292.1 XM_017011293.1 XM_017011294.1 XM_017011295.1 XM_017011296.1 XM_017011297.1 XM_017011298.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3253628 c.3373C>T p.Q1125* Chr.6: 31639520-31639520 on Build GRCh38
5457252 c.3190G>A p.A1064T Chr.6: 31639595-31639595 on Build GRCh38
1077620 c.3242G>A p.R1081Q Chr.6: 31639543-31639543 on Build GRCh38
4187949 c.3179C>A p.A1060D Chr.6: 31639606-31639606 on Build GRCh38
5457253 c.3298G>A p.A1100T Chr.6: 31639595-31639595 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776043632 Chr.6: 31639580-31639580 on Build GRCh38
rs771425695 Chr.6: 31639489-31639489 on Build GRCh38
rs756727682 Chr.6: 31639572-31639572 on Build GRCh38
rs763307225 Chr.6: 31639527-31639527 on Build GRCh38
rs775849509 Chr.6: 31639526-31639526 on Build GRCh38
rs768552143 Chr.6: 31639488-31639488 on Build GRCh38
rs993104131 Chr.6: 31639637-31639637 on Build GRCh38
rs148136430 Chr.6: 31639644-31639644 on Build GRCh38
rs776604391 Chr.6: 31639595-31639595 on Build GRCh38
rs764222954 Chr.6: 31639671-31639671 on Build GRCh38
rs200917529 Chr.6: 31639645-31639645 on Build GRCh38
rs753778566 Chr.6: 31639546-31639546 on Build GRCh38
rs746361226 Chr.6: 31639635-31639635 on Build GRCh38
rs547612475 Chr.6: 31639621-31639621 on Build GRCh38
rs749607453 Chr.6: 31639657-31639657 on Build GRCh38
rs751639269 Chr.6: 31639544-31639544 on Build GRCh38
rs778724129 Chr.6: 31639553-31639553 on Build GRCh38
rs746547600 Chr.6: 31639484-31639484 on Build GRCh38
rs981142289 Chr.6: 31639669-31639669 on Build GRCh38
rs767561330 Chr.6: 31639591-31639591 on Build GRCh38
rs113756104 Chr.6: 31639499-31639499 on Build GRCh38
rs749317845 Chr.6: 31639605-31639605 on Build GRCh38
rs773119841 Chr.6: 31639668-31639668 on Build GRCh38
rs770252601 Chr.6: 31639578-31639578 on Build GRCh38
rs749692025 Chr.6: 31639581-31639581 on Build GRCh38
rs532488148 Chr.6: 31639545-31639545 on Build GRCh38
rs746427524 Chr.6: 31639574-31639574 on Build GRCh38
rs546145122 Chr.6: 31639656-31639657 on Build GRCh38
rs761939891 Chr.6: 31639587-31639587 on Build GRCh38
rs765088766 Chr.6: 31639679-31639679 on Build GRCh38
rs200265396 Chr.6: 31639542-31639542 on Build GRCh38
rs202237524 Chr.6: 31639661-31639661 on Build GRCh38
rs970248636 Chr.6: 31639656-31639656 on Build GRCh38
rs566045731 Chr.6: 31639641-31639641 on Build GRCh38
rs570410084 Chr.6: 31639616-31639616 on Build GRCh38
rs769201593 Chr.6: 31639658-31639658 on Build GRCh38
rs754799151 Chr.6: 31639552-31639552 on Build GRCh38
rs771543249 Chr.6: 31639475-31639475 on Build GRCh38
rs369043420 Chr.6: 31639520-31639520 on Build GRCh38
rs1028445572 Chr.6: 31639482-31639482 on Build GRCh38
rs751225360 Chr.6: 31639622-31639622 on Build GRCh38
rs780606550 Chr.6: 31639631-31639631 on Build GRCh38
rs765184564 Chr.6: 31639597-31639597 on Build GRCh38
rs772171866 Chr.6: 31639666-31639666 on Build GRCh38
rs781712151 Chr.6: 31639573-31639573 on Build GRCh38
rs774730487 Chr.6: 31639586-31639586 on Build GRCh38
rs746459987 Chr.6: 31639567-31639567 on Build GRCh38
rs973420685 Chr.6: 31639582-31639582 on Build GRCh38
rs758000851 Chr.6: 31639563-31639563 on Build GRCh38
rs760843472 Chr.6: 31639675-31639675 on Build GRCh38
rs111986400 Chr.6: 31639497-31639497 on Build GRCh38
rs201690317 Chr.6: 31639480-31639480 on Build GRCh38
rs752328918 Chr.6: 31639557-31639557 on Build GRCh38
rs200133183 Chr.6: 31639629-31639629 on Build GRCh38
rs146864834 Chr.6: 31639564-31639564 on Build GRCh38
rs145840472 Chr.6: 31639598-31639598 on Build GRCh38

View Full Product Details