Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 50569224-50569465 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNA1G
Entrez Gene ID
8913
Gene Name
calcium voltage-gated channel subunit alpha1 G
Gene Aliases
Ca(V)T.1, Cav3.1, NBR13, SCA42
UniGene
Hs.591169
Gene Chromosome Location
Chr.17: 50560929-50629625 on Build GRCh38
Transcripts
NM_001256324.1 NM_001256325.1 NM_001256326.1 NM_001256327.1 NM_001256328.1 NM_001256329.1 NM_001256330.1 NM_001256331.1 NM_001256332.1 NM_001256333.1 NM_001256334.1 NM_001256359.1 NM_001256360.1 NM_001256361.1 NM_018896.4 NM_198376.2 NM_198377.2 NM_198378.2 NM_198379.2 NM_198380.2 NM_198382.2 NM_198383.2 NM_198384.2 NM_198385.2 NM_198386.2 NM_198387.2 NM_198388.2 NM_198396.2 NR_046054.1 NR_046055.1 NR_046056.1 NR_046057.1 NR_046058.1 XM_006722160.3 XM_006722161.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1384266 c.483C>T p.I161I Chr.17: 50569293-50569293 on Build GRCh38
436866 c.420C>G p.G140G Chr.17: 50569230-50569230 on Build GRCh38
4067675 c.460C>T p.R154W Chr.17: 50569270-50569270 on Build GRCh38
436865 c.420C>G p.G140G Chr.17: 50569230-50569230 on Build GRCh38
5824584 c.484G>A p.A162T Chr.17: 50569294-50569294 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs188827702 Chr.17: 50569386-50569386 on Build GRCh38
rs747761401 Chr.17: 50569313-50569313 on Build GRCh38
rs886772966 Chr.17: 50569364-50569364 on Build GRCh38
rs184328705 Chr.17: 50569288-50569288 on Build GRCh38
rs554053128 Chr.17: 50569261-50569261 on Build GRCh38
rs866881319 Chr.17: 50569266-50569266 on Build GRCh38
rs766407944 Chr.17: 50569328-50569328 on Build GRCh38
rs386797601 Chr.17: 50569441-50569442 on Build GRCh38
rs978061518 Chr.17: 50569401-50569401 on Build GRCh38
rs368561457 Chr.17: 50569291-50569291 on Build GRCh38
rs1025846965 Chr.17: 50569352-50569352 on Build GRCh38
rs1017184075 Chr.17: 50569442-50569442 on Build GRCh38
rs953895476 Chr.17: 50569318-50569318 on Build GRCh38
rs922049729 Chr.17: 50569441-50569441 on Build GRCh38
rs966964237 Chr.17: 50569393-50569393 on Build GRCh38
rs997950625 Chr.17: 50569351-50569351 on Build GRCh38
rs1039110654 Chr.17: 50569438-50569438 on Build GRCh38
rs773043628 Chr.17: 50569323-50569323 on Build GRCh38
rs768036026 Chr.17: 50569301-50569301 on Build GRCh38
rs372851469 Chr.17: 50569293-50569293 on Build GRCh38
rs375439167 Chr.17: 50569324-50569324 on Build GRCh38
rs749835211 Chr.17: 50569287-50569287 on Build GRCh38
rs936684694 Chr.17: 50569421-50569421 on Build GRCh38
rs556506413 Chr.17: 50569400-50569400 on Build GRCh38
rs769603658 Chr.17: 50569320-50569320 on Build GRCh38
rs925255497 Chr.17: 50569415-50569415 on Build GRCh38
rs576070172 Chr.17: 50569444-50569444 on Build GRCh38
rs754234255 Chr.17: 50569372-50569372 on Build GRCh38
rs900909619 Chr.17: 50569350-50569350 on Build GRCh38
rs776352826 Chr.17: 50569307-50569307 on Build GRCh38
rs759677534 Chr.17: 50569343-50569343 on Build GRCh38
rs567659601 Chr.17: 50569358-50569358 on Build GRCh38
rs774144486 Chr.17: 50569340-50569340 on Build GRCh38
rs1034544772 Chr.17: 50569308-50569308 on Build GRCh38
rs762860094 Chr.17: 50569270-50569270 on Build GRCh38

View Full Product Details