Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1200535-1200808 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNA1H
Entrez Gene ID
8912
Gene Name
calcium voltage-gated channel subunit alpha1 H
Gene Aliases
CACNA1HB, Cav3.2, ECA6, EIG6
UniGene
Hs.459642
Gene Chromosome Location
Chr.16: 1153241-1221772 on Build GRCh38
Transcripts
NM_001005407.1 NM_021098.2 XM_005255652.4 XM_006720963.3 XM_006720964.3 XM_006720965.3 XM_006720967.3 XM_006720968.3 XM_011522724.2 XM_011522727.2 XM_017023819.1 XM_017023820.1 XM_017023821.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
967071 c.1092C>T p.I364I Chr.16: 1200544-1200544 on Build GRCh38
2142846 c.1098C>T p.Y366Y Chr.16: 1200550-1200550 on Build GRCh38
967074 c.1095C>T p.G365G Chr.16: 1200547-1200547 on Build GRCh38
6582849 c.1119+2T>G p.? Chr.16: 1200573-1200573 on Build GRCh38
4255531 c.1098C>T p.Y366Y Chr.16: 1200550-1200550 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs973334540 Chr.16: 1200757-1200757 on Build GRCh38
rs1033347519 Chr.16: 1200690-1200690 on Build GRCh38
rs567920562 Chr.16: 1200641-1200641 on Build GRCh38
rs753602746 Chr.16: 1200621-1200621 on Build GRCh38
rs979970008 Chr.16: 1200663-1200663 on Build GRCh38
rs2659775 Chr.16: 1200611-1200611 on Build GRCh38
rs374385036 Chr.16: 1200588-1200588 on Build GRCh38
rs778810051 Chr.16: 1200667-1200667 on Build GRCh38
rs752699192 Chr.16: 1200669-1200669 on Build GRCh38
rs41292281 Chr.16: 1200763-1200763 on Build GRCh38
rs772848426 Chr.16: 1200578-1200578 on Build GRCh38
rs770416013 Chr.16: 1200773-1200773 on Build GRCh38
rs779946212 Chr.16: 1200553-1200553 on Build GRCh38
rs754903643 Chr.16: 1200564-1200564 on Build GRCh38
rs571309152 Chr.16: 1200662-1200663 on Build GRCh38
rs866199752 Chr.16: 1200560-1200560 on Build GRCh38
rs989690921 Chr.16: 1200617-1200617 on Build GRCh38
rs71278995 Chr.16: 1200608-1200611 on Build GRCh38
rs777405180 Chr.16: 1200706-1200707 on Build GRCh38
rs943269151 Chr.16: 1200642-1200642 on Build GRCh38
rs766565300 Chr.16: 1200709-1200709 on Build GRCh38
rs375934557 Chr.16: 1200642-1200642 on Build GRCh38
rs773385518 Chr.16: 1200705-1200705 on Build GRCh38
rs879842927 Chr.16: 1200629-1200629 on Build GRCh38
rs999691704 Chr.16: 1200635-1200635 on Build GRCh38
rs554412075 Chr.16: 1200634-1200634 on Build GRCh38
rs150200429 Chr.16: 1200694-1200694 on Build GRCh38
rs878880304 Chr.16: 1200627-1200627 on Build GRCh38
rs780717105 Chr.16: 1200686-1200686 on Build GRCh38
rs777317154 Chr.16: 1200680-1200680 on Build GRCh38
rs750648909 Chr.16: 1200614-1200614 on Build GRCh38
rs368268122 Chr.16: 1200704-1200704 on Build GRCh38
rs8044363 Chr.16: 1200559-1200559 on Build GRCh38
rs369185990 Chr.16: 1200754-1200754 on Build GRCh38
rs2939840 Chr.16: 1200664-1200664 on Build GRCh38
rs761975796 Chr.16: 1200610-1200610 on Build GRCh38
rs766540396 Chr.16: 1200620-1200620 on Build GRCh38
rs766868659 Chr.16: 1200634-1200635 on Build GRCh38
rs992888533 Chr.16: 1200646-1200646 on Build GRCh38
rs985503035 Chr.16: 1200739-1200739 on Build GRCh38
rs1037587280 Chr.16: 1200653-1200653 on Build GRCh38
rs770036532 Chr.16: 1200593-1200593 on Build GRCh38
rs768965884 Chr.16: 1200692-1200692 on Build GRCh38
rs1031209556 Chr.16: 1200640-1200640 on Build GRCh38
rs906359032 Chr.16: 1200774-1200774 on Build GRCh38
rs767742306 Chr.16: 1200612-1200612 on Build GRCh38
rs575226771 Chr.16: 1200687-1200687 on Build GRCh38
rs772638826 Chr.16: 1200758-1200758 on Build GRCh38
rs1049787128 Chr.16: 1200637-1200637 on Build GRCh38
rs59091981 Chr.16: 1200708-1200708 on Build GRCh38
rs753249004 Chr.16: 1200618-1200619 on Build GRCh38
rs747600250 Chr.16: 1200574-1200574 on Build GRCh38
rs756579097 Chr.16: 1200621-1200621 on Build GRCh38
rs775878707 Chr.16: 1200594-1200594 on Build GRCh38
rs529122240 Chr.16: 1200725-1200725 on Build GRCh38
rs1047764685 Chr.16: 1200561-1200561 on Build GRCh38
rs917025676 Chr.16: 1200659-1200659 on Build GRCh38
rs762456183 Chr.16: 1200702-1200702 on Build GRCh38
rs1022855888 Chr.16: 1200633-1200633 on Build GRCh38
rs536962880 Chr.16: 1200662-1200662 on Build GRCh38
rs757874585 Chr.16: 1200675-1200675 on Build GRCh38
rs769117669 Chr.16: 1200752-1200752 on Build GRCh38
rs770983743 Chr.16: 1200768-1200768 on Build GRCh38
rs760796172 Chr.16: 1200766-1200766 on Build GRCh38
rs188397970 Chr.16: 1200711-1200711 on Build GRCh38
rs978309224 Chr.16: 1200707-1200707 on Build GRCh38
rs898958342 Chr.16: 1200563-1200563 on Build GRCh38
rs751019130 Chr.16: 1200733-1200733 on Build GRCh38
rs748730819 Chr.16: 1200707-1200707 on Build GRCh38
rs372159580 Chr.16: 1200577-1200577 on Build GRCh38
rs760900511 Chr.16: 1200616-1200616 on Build GRCh38
rs758285252 Chr.16: 1200723-1200723 on Build GRCh38
rs1037578858 Chr.16: 1200562-1200562 on Build GRCh38
rs2939839 Chr.16: 1200657-1200657 on Build GRCh38
rs764598327 Chr.16: 1200609-1200609 on Build GRCh38
rs944746813 Chr.16: 1200700-1200700 on Build GRCh38
rs202181749 Chr.16: 1200599-1200599 on Build GRCh38
rs748551891 Chr.16: 1200755-1200755 on Build GRCh38
rs996377281 Chr.16: 1200628-1200628 on Build GRCh38
rs746527382 Chr.16: 1200682-1200682 on Build GRCh38
rs765058001 Chr.16: 1200712-1200712 on Build GRCh38
rs559931034 Chr.16: 1200713-1200713 on Build GRCh38
rs745543078 Chr.16: 1200751-1200751 on Build GRCh38
rs377726477 Chr.16: 1200638-1200638 on Build GRCh38
rs375121203 Chr.16: 1200740-1200740 on Build GRCh38
rs201765670 Chr.16: 1200683-1200683 on Build GRCh38
rs778826216 Chr.16: 1200565-1200565 on Build GRCh38
rs781498544 Chr.16: 1200580-1200580 on Build GRCh38
rs934051068 Chr.16: 1200635-1200635 on Build GRCh38
rs2659773 Chr.16: 1200608-1200608 on Build GRCh38
rs780889215 Chr.16: 1200743-1200743 on Build GRCh38
rs2659772 Chr.16: 1200574-1200574 on Build GRCh38
rs939109422 Chr.16: 1200775-1200775 on Build GRCh38
rs932024389 Chr.16: 1200738-1200738 on Build GRCh38
rs904067921 Chr.16: 1200772-1200772 on Build GRCh38

View Full Product Details