Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 37274676-37274921 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CELF4
Entrez Gene ID
56853
Gene Name
CUGBP, Elav-like family member 4
Gene Aliases
BRUNOL4, CELF-4
UniGene
Hs.435976
Gene Chromosome Location
Chr.18: 37243040-37566052 on Build GRCh38
Transcripts
NM_001025087.1 NM_001025088.1 NM_001025089.1 NM_020180.3 XM_005258308.3 XM_011526079.2 XM_011526081.2 XM_011526083.2 XM_011526084.2 XM_011526085.2 XM_011526086.2 XM_011526087.2 XM_011526090.2 XM_011526091.2 XM_011526092.2 XM_017025840.1 XM_017025841.1 XM_017025842.1 XM_017025843.1 XM_017025844.1 XM_017025845.1 XM_017025846.1 XM_017025847.1 XM_017025848.1 XM_017025849.1 XM_017025850.1 XM_017025851.1 XM_017025852.1 XM_017025853.1 XM_017025854.1 XM_017025855.1 XM_017025856.1 XM_017025857.1 XM_017025858.1 XM_017025859.1 XM_017025860.1 XM_017025861.1 XM_017025862.1 XM_017025863.1 XM_017025864.1 XM_017025865.1 XM_017025866.1 XM_017025867.1 XM_017025868.1 XM_017025869.1 XM_017025870.1 XM_017025871.1 XM_017025872.1 NR_134588.1

Gene Symbol
LOC105372068
Entrez Gene ID
105372068
Gene Name
uncharacterized LOC105372068
Gene Aliases
-
UniGene
Hs.569903
Gene Chromosome Location
Chr.18: 37274460-37276400 on Build GRCh38
Transcripts
NM_001025087.1 NM_001025088.1 NM_001025089.1 NM_020180.3 XM_005258308.3 XM_011526079.2 XM_011526081.2 XM_011526083.2 XM_011526084.2 XM_011526085.2 XM_011526086.2 XM_011526087.2 XM_011526090.2 XM_011526091.2 XM_011526092.2 XM_017025840.1 XM_017025841.1 XM_017025842.1 XM_017025843.1 XM_017025844.1 XM_017025845.1 XM_017025846.1 XM_017025847.1 XM_017025848.1 XM_017025849.1 XM_017025850.1 XM_017025851.1 XM_017025852.1 XM_017025853.1 XM_017025854.1 XM_017025855.1 XM_017025856.1 XM_017025857.1 XM_017025858.1 XM_017025859.1 XM_017025860.1 XM_017025861.1 XM_017025862.1 XM_017025863.1 XM_017025864.1 XM_017025865.1 XM_017025866.1 XM_017025867.1 XM_017025868.1 XM_017025869.1 XM_017025870.1 XM_017025871.1 XM_017025872.1 NR_134588.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5748949 c.640G>A p.G214S Chr.18: 37274822-37274822 on Build GRCh38
6431647 c.614C>T p.A205V Chr.18: 37274848-37274848 on Build GRCh38
5774658 c.621C>T p.A207A Chr.18: 37274841-37274841 on Build GRCh38
3692114 c.622G>A p.A208T Chr.18: 37274840-37274840 on Build GRCh38
6532649 c.651C>G p.T217T Chr.18: 37274811-37274811 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370765225 Chr.18: 37274865-37274865 on Build GRCh38
rs752215681 Chr.18: 37274798-37274798 on Build GRCh38
rs192146200 Chr.18: 37274832-37274832 on Build GRCh38
rs760050764 Chr.18: 37274789-37274789 on Build GRCh38
rs762086455 Chr.18: 37274740-37274740 on Build GRCh38
rs781145626 Chr.18: 37274853-37274853 on Build GRCh38
rs1048645854 Chr.18: 37274770-37274770 on Build GRCh38
rs373551622 Chr.18: 37274757-37274757 on Build GRCh38
rs540241193 Chr.18: 37274895-37274895 on Build GRCh38
rs772310513 Chr.18: 37274904-37274904 on Build GRCh38
rs770959900 Chr.18: 37274731-37274731 on Build GRCh38
rs1016554729 Chr.18: 37274751-37274751 on Build GRCh38
rs998063739 Chr.18: 37274901-37274901 on Build GRCh38
rs764738264 Chr.18: 37274790-37274790 on Build GRCh38
rs918295372 Chr.18: 37274752-37274752 on Build GRCh38
rs774295794 Chr.18: 37274738-37274738 on Build GRCh38
rs1042050004 Chr.18: 37274792-37274792 on Build GRCh38
rs777610330 Chr.18: 37274699-37274699 on Build GRCh38
rs914959759 Chr.18: 37274781-37274781 on Build GRCh38
rs746828051 Chr.18: 37274716-37274716 on Build GRCh38
rs928099632 Chr.18: 37274717-37274717 on Build GRCh38
rs533853605 Chr.18: 37274899-37274899 on Build GRCh38
rs751212038 Chr.18: 37274775-37274776 on Build GRCh38
rs199540513 Chr.18: 37274806-37274806 on Build GRCh38
rs766762883 Chr.18: 37274768-37274768 on Build GRCh38
rs777330008 Chr.18: 37274826-37274826 on Build GRCh38
rs942418044 Chr.18: 37274797-37274797 on Build GRCh38
rs745819955 Chr.18: 37274736-37274736 on Build GRCh38
rs972276457 Chr.18: 37274745-37274745 on Build GRCh38
rs928102767 Chr.18: 37274754-37274754 on Build GRCh38
rs761070723 Chr.18: 37274759-37274759 on Build GRCh38
rs937815188 Chr.18: 37274755-37274755 on Build GRCh38
rs900413033 Chr.18: 37274756-37274756 on Build GRCh38
rs772244747 Chr.18: 37274748-37274748 on Build GRCh38
rs756940136 Chr.18: 37274778-37274778 on Build GRCh38
rs757067390 Chr.18: 37274841-37274841 on Build GRCh38
rs973337590 Chr.18: 37274732-37274732 on Build GRCh38
rs745729950 Chr.18: 37274862-37274862 on Build GRCh38
rs554591753 Chr.18: 37274735-37274735 on Build GRCh38
rs758892352 Chr.18: 37274894-37274894 on Build GRCh38
rs376106133 Chr.18: 37274783-37274783 on Build GRCh38
rs996448349 Chr.18: 37274788-37274788 on Build GRCh38
rs865904398 Chr.18: 37274808-37274808 on Build GRCh38

View Full Product Details