Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 56182083-56182340 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALP
Entrez Gene ID
85569
Gene Name
galanin like peptide
Gene Aliases
-
UniGene
Hs.283915
Gene Chromosome Location
Chr.19: 56176020-56185775 on Build GRCh38
Transcripts
NM_001145546.1 NM_033106.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1712923 c.169G>A p.G57R Chr.19: 56182204-56182204 on Build GRCh38
7115861 c.212C>A p.A71D Chr.19: 56182247-56182247 on Build GRCh38
3713178 c.139C>A p.L47I Chr.19: 56182174-56182174 on Build GRCh38
7097263 c.166G>T p.D56Y Chr.19: 56182201-56182201 on Build GRCh38
3539922 c.211G>A p.A71T Chr.19: 56182246-56182246 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751296909 Chr.19: 56182199-56182199 on Build GRCh38
rs368664274 Chr.19: 56182136-56182136 on Build GRCh38
rs753750615 Chr.19: 56182259-56182259 on Build GRCh38
rs570045014 Chr.19: 56182294-56182294 on Build GRCh38
rs200545163 Chr.19: 56182168-56182168 on Build GRCh38
rs757877490 Chr.19: 56182264-56182264 on Build GRCh38
rs539516717 Chr.19: 56182128-56182128 on Build GRCh38
rs182015294 Chr.19: 56182190-56182190 on Build GRCh38
rs758645278 Chr.19: 56182142-56182142 on Build GRCh38
rs1038324260 Chr.19: 56182112-56182112 on Build GRCh38
rs145594097 Chr.19: 56182200-56182200 on Build GRCh38
rs201851941 Chr.19: 56182156-56182156 on Build GRCh38
rs777031197 Chr.19: 56182195-56182195 on Build GRCh38
rs775869241 Chr.19: 56182297-56182297 on Build GRCh38
rs36004192 Chr.19: 56182193-56182193 on Build GRCh38
rs274174 Chr.19: 56182274-56182274 on Build GRCh38
rs774610045 Chr.19: 56182299-56182299 on Build GRCh38
rs367649834 Chr.19: 56182302-56182302 on Build GRCh38
rs766242217 Chr.19: 56182258-56182258 on Build GRCh38
rs758515513 Chr.19: 56182144-56182144 on Build GRCh38
rs771308137 Chr.19: 56182194-56182194 on Build GRCh38
rs757198597 Chr.19: 56182273-56182277 on Build GRCh38
rs1015004378 Chr.19: 56182233-56182233 on Build GRCh38
rs773611664 Chr.19: 56182126-56182126 on Build GRCh38
rs200014398 Chr.19: 56182175-56182175 on Build GRCh38
rs750188898 Chr.19: 56182152-56182152 on Build GRCh38
rs769772853 Chr.19: 56182286-56182286 on Build GRCh38
rs752761393 Chr.19: 56182137-56182137 on Build GRCh38
rs202229314 Chr.19: 56182177-56182177 on Build GRCh38
rs770611174 Chr.19: 56182275-56182275 on Build GRCh38
rs3745833 Chr.19: 56182251-56182251 on Build GRCh38
rs778728668 Chr.19: 56182263-56182263 on Build GRCh38
rs772854458 Chr.19: 56182189-56182189 on Build GRCh38
rs371254878 Chr.19: 56182262-56182262 on Build GRCh38
rs768512469 Chr.19: 56182169-56182169 on Build GRCh38
rs754374071 Chr.19: 56182130-56182130 on Build GRCh38
rs200881395 Chr.19: 56182188-56182188 on Build GRCh38
rs762960139 Chr.19: 56182211-56182211 on Build GRCh38
rs766805456 Chr.19: 56182129-56182129 on Build GRCh38
rs980205762 Chr.19: 56182296-56182296 on Build GRCh38
rs377761776 Chr.19: 56182203-56182203 on Build GRCh38
rs751204520 Chr.19: 56182213-56182213 on Build GRCh38
rs956194473 Chr.19: 56182215-56182215 on Build GRCh38
rs770280916 Chr.19: 56182287-56182287 on Build GRCh38
rs373873063 Chr.19: 56182139-56182139 on Build GRCh38
rs747668035 Chr.19: 56182184-56182184 on Build GRCh38
rs374359432 Chr.19: 56182266-56182266 on Build GRCh38
rs752811117 Chr.19: 56182204-56182204 on Build GRCh38
rs145230182 Chr.19: 56182298-56182298 on Build GRCh38
rs755981643 Chr.19: 56182155-56182155 on Build GRCh38
rs759564236 Chr.19: 56182133-56182133 on Build GRCh38

View Full Product Details