Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 55350903-55351152 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO34
Entrez Gene ID
55030
Gene Name
F-box protein 34
Gene Aliases
CGI-301, Fbx34
UniGene
Hs.525348
Gene Chromosome Location
Chr.14: 55271303-55355222 on Build GRCh38
Transcripts
NM_017943.3 NM_152231.1 XM_005267791.4 XM_006720185.3 XM_017021391.1 XM_017021392.1 XM_017021393.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3814955 c.713T>G p.F238C Chr.14: 55351103-55351103 on Build GRCh38
2245320 c.667G>C p.A223P Chr.14: 55351057-55351057 on Build GRCh38
6662209 c.664C>T p.L222L Chr.14: 55351054-55351054 on Build GRCh38
3814954 c.679A>C p.K227Q Chr.14: 55351069-55351069 on Build GRCh38
470040 c.531T>G p.P177P Chr.14: 55350921-55350921 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371770836 Chr.14: 55350950-55350950 on Build GRCh38
rs879833377 Chr.14: 55351040-55351040 on Build GRCh38
rs199674550 Chr.14: 55351068-55351068 on Build GRCh38
rs753567490 Chr.14: 55350995-55350995 on Build GRCh38
rs773399959 Chr.14: 55351013-55351013 on Build GRCh38
rs199521219 Chr.14: 55350932-55350932 on Build GRCh38
rs200456651 Chr.14: 55351046-55351046 on Build GRCh38
rs147933450 Chr.14: 55351074-55351074 on Build GRCh38
rs775251680 Chr.14: 55351077-55351077 on Build GRCh38
rs749433625 Chr.14: 55351043-55351043 on Build GRCh38
rs752692331 Chr.14: 55350982-55350982 on Build GRCh38
rs748717813 Chr.14: 55351000-55351000 on Build GRCh38
rs1008347470 Chr.14: 55351063-55351063 on Build GRCh38
rs750105888 Chr.14: 55351120-55351120 on Build GRCh38
rs770461440 Chr.14: 55351003-55351003 on Build GRCh38
rs767289548 Chr.14: 55350968-55350968 on Build GRCh38
rs755376786 Chr.14: 55350996-55350996 on Build GRCh38
rs764067393 Chr.14: 55351087-55351087 on Build GRCh38
rs747334974 Chr.14: 55350994-55350994 on Build GRCh38
rs756831688 Chr.14: 55351096-55351096 on Build GRCh38
rs759192782 Chr.14: 55351065-55351065 on Build GRCh38
rs763233001 Chr.14: 55350931-55350931 on Build GRCh38
rs755531600 Chr.14: 55350983-55350983 on Build GRCh38
rs143781588 Chr.14: 55351082-55351082 on Build GRCh38
rs778599320 Chr.14: 55350992-55350992 on Build GRCh38
rs142017596 Chr.14: 55351093-55351093 on Build GRCh38
rs541406946 Chr.14: 55351061-55351061 on Build GRCh38
rs753386438 Chr.14: 55350989-55350989 on Build GRCh38
rs930774185 Chr.14: 55351060-55351060 on Build GRCh38
rs145907652 Chr.14: 55351113-55351113 on Build GRCh38
rs763627716 Chr.14: 55350984-55350984 on Build GRCh38
rs773524764 Chr.14: 55350930-55350930 on Build GRCh38
rs10144418 Chr.14: 55350990-55350990 on Build GRCh38
rs138652671 Chr.14: 55351115-55351115 on Build GRCh38
rs769933483 Chr.14: 55350928-55350928 on Build GRCh38
rs889911386 Chr.14: 55351062-55351062 on Build GRCh38
rs372775034 Chr.14: 55351012-55351012 on Build GRCh38
rs774274718 Chr.14: 55350943-55350943 on Build GRCh38

View Full Product Details