Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1716263-1716536 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK11A
Entrez Gene ID
728642
Gene Name
cyclin dependent kinase 11A
Gene Aliases
CDC2L2, CDC2L3, CDK11-p110, CDK11-p46, CDK11-p58, PITSLRE, p58GTA
UniGene
Hs.651228
Gene Chromosome Location
Chr.1: 1702383-1724565 on Build GRCh38
Transcripts
NM_001313896.1 NM_001313982.1 NM_024011.3 NM_033529.3 NR_132739.1 NR_132740.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4224752 c.350G>A p.R117K Chr.1: 1716454-1716454 on Build GRCh38
898874 c.346G>T p.E116* Chr.1: 1716458-1716458 on Build GRCh38
424451 c.452G>A p.R151K Chr.1: 1716352-1716352 on Build GRCh38
6625325 c.401G>A p.R134Q Chr.1: 1716403-1716403 on Build GRCh38
5446030 c.342_343insAAAGAA p.E116_R117insKE Chr.1: 1716461-1716462 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370303160 Chr.1: 1716341-1716341 on Build GRCh38
rs745508835 Chr.1: 1716352-1716352 on Build GRCh38
rs542594220 Chr.1: 1716401-1716401 on Build GRCh38
rs74045984 Chr.1: 1716314-1716314 on Build GRCh38
rs866937199 Chr.1: 1716462-1716462 on Build GRCh38
rs760989414 Chr.1: 1716378-1716378 on Build GRCh38
rs767944042 Chr.1: 1716458-1716459 on Build GRCh38
rs746108190 Chr.1: 1716477-1716477 on Build GRCh38
rs200272503 Chr.1: 1716404-1716404 on Build GRCh38
rs753458825 Chr.1: 1716407-1716407 on Build GRCh38
rs745434920 Chr.1: 1716313-1716313 on Build GRCh38
rs386627543 Chr.1: 1716500-1716502 on Build GRCh38
rs201839990 Chr.1: 1716501-1716502 on Build GRCh38
rs754630865 Chr.1: 1716447-1716447 on Build GRCh38
rs775549112 Chr.1: 1716483-1716483 on Build GRCh38
rs748675685 Chr.1: 1716365-1716365 on Build GRCh38
rs746739655 Chr.1: 1716297-1716297 on Build GRCh38
rs766098874 Chr.1: 1716442-1716442 on Build GRCh38
rs796229119 Chr.1: 1716291-1716291 on Build GRCh38
rs760387372 Chr.1: 1716440-1716440 on Build GRCh38
rs750242434 Chr.1: 1716439-1716439 on Build GRCh38
rs760718586 Chr.1: 1716338-1716338 on Build GRCh38
rs776712506 Chr.1: 1716427-1716427 on Build GRCh38
rs762257977 Chr.1: 1716332-1716332 on Build GRCh38
rs780620408 Chr.1: 1716499-1716503 on Build GRCh38
rs747279722 Chr.1: 1716424-1716424 on Build GRCh38
rs757040406 Chr.1: 1716347-1716347 on Build GRCh38
rs769000712 Chr.1: 1716326-1716326 on Build GRCh38
rs199846682 Chr.1: 1716498-1716499 on Build GRCh38
rs144636354 Chr.1: 1716454-1716455 on Build GRCh38
rs762479182 Chr.1: 1716456-1716457 on Build GRCh38
rs769960600 Chr.1: 1716479-1716479 on Build GRCh38
rs750982385 Chr.1: 1716492-1716495 on Build GRCh38
rs752188532 Chr.1: 1716450-1716450 on Build GRCh38
rs756915742 Chr.1: 1716305-1716305 on Build GRCh38
rs751412468 Chr.1: 1716346-1716346 on Build GRCh38
rs199510897 Chr.1: 1716434-1716434 on Build GRCh38
rs772118548 Chr.1: 1716495-1716495 on Build GRCh38
rs199866927 Chr.1: 1716461-1716462 on Build GRCh38
rs1883424 Chr.1: 1716489-1716489 on Build GRCh38
rs1051874119 Chr.1: 1716452-1716453 on Build GRCh38
rs183746110 Chr.1: 1716343-1716343 on Build GRCh38
rs200132561 Chr.1: 1716508-1716508 on Build GRCh38
rs368053496 Chr.1: 1716405-1716405 on Build GRCh38
rs72901775 Chr.1: 1716375-1716375 on Build GRCh38
rs752313298 Chr.1: 1716510-1716510 on Build GRCh38
rs200224067 Chr.1: 1716454-1716454 on Build GRCh38
rs779642694 Chr.1: 1716358-1716358 on Build GRCh38
rs766112607 Chr.1: 1716502-1716502 on Build GRCh38
rs570927461 Chr.1: 1716422-1716422 on Build GRCh38
rs72901773 Chr.1: 1716339-1716339 on Build GRCh38
rs777576455 Chr.1: 1716296-1716296 on Build GRCh38
rs568553222 Chr.1: 1716505-1716505 on Build GRCh38
rs770810430 Chr.1: 1716450-1716451 on Build GRCh38
rs767371982 Chr.1: 1716435-1716435 on Build GRCh38
rs779200159 Chr.1: 1716332-1716332 on Build GRCh38
rs771243407 Chr.1: 1716426-1716426 on Build GRCh38
rs776785110 Chr.1: 1716380-1716380 on Build GRCh38
rs769348783 Chr.1: 1716371-1716371 on Build GRCh38
rs767920717 Chr.1: 1716333-1716333 on Build GRCh38
rs900168817 Chr.1: 1716387-1716387 on Build GRCh38
rs772544700 Chr.1: 1716373-1716373 on Build GRCh38
rs771913487 Chr.1: 1716361-1716362 on Build GRCh38
rs201054761 Chr.1: 1716419-1716419 on Build GRCh38
rs370418133 Chr.1: 1716406-1716406 on Build GRCh38
rs759520770 Chr.1: 1716389-1716389 on Build GRCh38
rs72634830 Chr.1: 1716306-1716306 on Build GRCh38
rs775907923 Chr.1: 1716321-1716321 on Build GRCh38
rs200816491 Chr.1: 1716500-1716500 on Build GRCh38
rs773149307 Chr.1: 1716380-1716381 on Build GRCh38
rs760416247 Chr.1: 1716422-1716422 on Build GRCh38
rs771167585 Chr.1: 1716471-1716471 on Build GRCh38
rs112748730 Chr.1: 1716463-1716463 on Build GRCh38
rs755670050 Chr.1: 1716356-1716356 on Build GRCh38
rs374642406 Chr.1: 1716433-1716433 on Build GRCh38
rs774873773 Chr.1: 1716329-1716329 on Build GRCh38
rs748566008 Chr.1: 1716372-1716372 on Build GRCh38
rs531731523 Chr.1: 1716350-1716350 on Build GRCh38
rs748188287 Chr.1: 1716347-1716347 on Build GRCh38
rs772669213 Chr.1: 1716420-1716420 on Build GRCh38
rs776399480 Chr.1: 1716453-1716456 on Build GRCh38
rs796957281 Chr.1: 1716290-1716290 on Build GRCh38
rs200601104 Chr.1: 1716500-1716500 on Build GRCh38
rs772995447 Chr.1: 1716497-1716497 on Build GRCh38
rs72909014 Chr.1: 1716432-1716432 on Build GRCh38
rs754510372 Chr.1: 1716413-1716413 on Build GRCh38
rs749820169 Chr.1: 1716325-1716325 on Build GRCh38
rs371899640 Chr.1: 1716443-1716443 on Build GRCh38
rs778129644 Chr.1: 1716455-1716455 on Build GRCh38
rs751465521 Chr.1: 1716403-1716403 on Build GRCh38
rs527915790 Chr.1: 1716417-1716417 on Build GRCh38
rs773539187 Chr.1: 1716337-1716337 on Build GRCh38
rs759268317 Chr.1: 1716506-1716506 on Build GRCh38
rs200249552 Chr.1: 1716297-1716306 on Build GRCh38

View Full Product Details