Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 39846122-39846381 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBL
Entrez Gene ID
2091
Gene Name
fibrillarin
Gene Aliases
FIB, FLRN, RNU3IP1
UniGene
Hs.299002
Gene Chromosome Location
Chr.19: 39834453-39846414 on Build GRCh38
Transcripts
NM_001436.3 XM_005258651.2 XM_011526623.2

SNP Detail

SNP ID Chromosome Location
rs778942544 Chr.19: 39846201-39846201 on Build GRCh38
rs780152843 Chr.19: 39846323-39846323 on Build GRCh38
rs771659204 Chr.19: 39846342-39846342 on Build GRCh38
rs763824396 Chr.19: 39846352-39846352 on Build GRCh38
rs563820734 Chr.19: 39846235-39846235 on Build GRCh38
rs767202645 Chr.19: 39846280-39846280 on Build GRCh38
rs377465638 Chr.19: 39846281-39846281 on Build GRCh38
rs747291724 Chr.19: 39846245-39846245 on Build GRCh38
rs956975679 Chr.19: 39846150-39846150 on Build GRCh38
rs768876295 Chr.19: 39846246-39846246 on Build GRCh38
rs373111008 Chr.19: 39846335-39846335 on Build GRCh38
rs760180832 Chr.19: 39846351-39846351 on Build GRCh38
rs1030416292 Chr.19: 39846275-39846275 on Build GRCh38
rs749859989 Chr.19: 39846336-39846336 on Build GRCh38
rs969016165 Chr.19: 39846331-39846331 on Build GRCh38
rs903336713 Chr.19: 39846166-39846166 on Build GRCh38
rs896179039 Chr.19: 39846228-39846228 on Build GRCh38
rs201229179 Chr.19: 39846217-39846217 on Build GRCh38
rs200865220 Chr.19: 39846258-39846258 on Build GRCh38
rs368706410 Chr.19: 39846358-39846358 on Build GRCh38
rs528648044 Chr.19: 39846310-39846310 on Build GRCh38
rs923506752 Chr.19: 39846159-39846159 on Build GRCh38
rs999937417 Chr.19: 39846330-39846330 on Build GRCh38
rs377454089 Chr.19: 39846157-39846157 on Build GRCh38
rs370831969 Chr.19: 39846269-39846269 on Build GRCh38
rs575641672 Chr.19: 39846238-39846238 on Build GRCh38
rs775099415 Chr.19: 39846349-39846349 on Build GRCh38
rs533865694 Chr.19: 39846155-39846155 on Build GRCh38
rs1052170298 Chr.19: 39846343-39846343 on Build GRCh38
rs184643003 Chr.19: 39846270-39846270 on Build GRCh38
rs373225683 Chr.19: 39846234-39846234 on Build GRCh38
rs568646907 Chr.19: 39846333-39846333 on Build GRCh38
rs747148798 Chr.19: 39846329-39846329 on Build GRCh38
rs376533903 Chr.19: 39846282-39846282 on Build GRCh38
rs763757645 Chr.19: 39846301-39846301 on Build GRCh38
rs199915708 Chr.19: 39846318-39846318 on Build GRCh38
rs770428417 Chr.19: 39846266-39846266 on Build GRCh38
rs754993919 Chr.19: 39846142-39846142 on Build GRCh38
rs574897970 Chr.19: 39846189-39846189 on Build GRCh38
rs988276683 Chr.19: 39846359-39846359 on Build GRCh38
rs910648578 Chr.19: 39846215-39846215 on Build GRCh38
rs546898553 Chr.19: 39846315-39846315 on Build GRCh38
rs780254143 Chr.19: 39846243-39846243 on Build GRCh38
rs745590088 Chr.19: 39846276-39846276 on Build GRCh38
rs200033307 Chr.19: 39846317-39846317 on Build GRCh38
rs376833478 Chr.19: 39846354-39846354 on Build GRCh38
rs933328097 Chr.19: 39846185-39846185 on Build GRCh38
rs777073749 Chr.19: 39846252-39846252 on Build GRCh38
rs1016119829 Chr.19: 39846267-39846267 on Build GRCh38
rs555667331 Chr.19: 39846158-39846158 on Build GRCh38
rs760398600 Chr.19: 39846293-39846293 on Build GRCh38
rs73550482 Chr.19: 39846320-39846320 on Build GRCh38
rs370884137 Chr.19: 39846332-39846332 on Build GRCh38

View Full Product Details