Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 25012704-25012959 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARX
Entrez Gene ID
170302
Gene Name
aristaless related homeobox
Gene Aliases
CT121, EIEE1, ISSX, MRX29, MRX32, MRX33, MRX36, MRX38, MRX43, MRX54, MRX76, MRX87, MRXS1, PRTS
UniGene
Hs.300304
Gene Chromosome Location
Chr.X: 25003694-25015948 on Build GRCh38
Transcripts
NM_139058.2

SNP Detail

SNP ID Chromosome Location
rs755049856 Chr.X: 25012753-25012753 on Build GRCh38
rs767831085 Chr.X: 25012875-25012875 on Build GRCh38
rs934260140 Chr.X: 25012735-25012735 on Build GRCh38
rs757785751 Chr.X: 25012894-25012894 on Build GRCh38
rs905289933 Chr.X: 25012796-25012796 on Build GRCh38
rs779281432 Chr.X: 25012901-25012901 on Build GRCh38
rs756365044 Chr.X: 25012882-25012882 on Build GRCh38
rs753910687 Chr.X: 25012892-25012892 on Build GRCh38
rs952320530 Chr.X: 25012771-25012771 on Build GRCh38
rs746914590 Chr.X: 25012742-25012742 on Build GRCh38
rs73473563 Chr.X: 25012731-25012731 on Build GRCh38
rs764290914 Chr.X: 25012883-25012883 on Build GRCh38
rs1001362221 Chr.X: 25012821-25012821 on Build GRCh38
rs1026904152 Chr.X: 25012754-25012754 on Build GRCh38
rs923319698 Chr.X: 25012842-25012842 on Build GRCh38
rs1038439680 Chr.X: 25012795-25012795 on Build GRCh38
rs896924340 Chr.X: 25012729-25012729 on Build GRCh38
rs976074798 Chr.X: 25012835-25012836 on Build GRCh38
rs1014624198 Chr.X: 25012839-25012839 on Build GRCh38
rs776280666 Chr.X: 25012902-25012902 on Build GRCh38
rs768546815 Chr.X: 25012757-25012757 on Build GRCh38
rs398124505 Chr.X: 25012935-25012935 on Build GRCh38
rs779717915 Chr.X: 25012913-25012913 on Build GRCh38
rs752936593 Chr.X: 25012880-25012880 on Build GRCh38
rs1017548359 Chr.X: 25012833-25012834 on Build GRCh38
rs959984312 Chr.X: 25012833-25012833 on Build GRCh38
rs758664610 Chr.X: 25012908-25012908 on Build GRCh38
rs1034621188 Chr.X: 25012832-25012832 on Build GRCh38
rs746690813 Chr.X: 25012924-25012924 on Build GRCh38
rs1005563217 Chr.X: 25012782-25012782 on Build GRCh38
rs768243127 Chr.X: 25012925-25012925 on Build GRCh38
rs1020167290 Chr.X: 25012845-25012845 on Build GRCh38
rs104894743 Chr.X: 25012937-25012937 on Build GRCh38
rs762645204 Chr.X: 25012872-25012872 on Build GRCh38

View Full Product Details