Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 97657911-97658140 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTR1B
Entrez Gene ID
10120
Gene Name
ARP1 actin-related protein 1 homolog B, centractin beta
Gene Aliases
ARP1B, CTRN2, PC3
UniGene
Hs.98791
Gene Chromosome Location
Chr.2: 97655939-97664098 on Build GRCh38
Transcripts
NM_005735.3 XM_005263854.4 XM_017003116.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7238367 c.840C>T p.A280A Chr.2: 97658028-97658028 on Build GRCh38
6593083 c.763C>T p.R255* Chr.2: 97658105-97658105 on Build GRCh38
384378 c.813G>C p.E271D Chr.2: 97658055-97658055 on Build GRCh38
5863042 c.898C>T p.L300F Chr.2: 97657970-97657970 on Build GRCh38
4096458 c.862A>G p.M288V Chr.2: 97658006-97658006 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138142670 Chr.2: 97657982-97657982 on Build GRCh38
rs368032697 Chr.2: 97657974-97657974 on Build GRCh38
rs753127424 Chr.2: 97657997-97657997 on Build GRCh38
rs769565201 Chr.2: 97658034-97658034 on Build GRCh38
rs777679337 Chr.2: 97658009-97658009 on Build GRCh38
rs1026006041 Chr.2: 97658006-97658006 on Build GRCh38
rs751531607 Chr.2: 97658098-97658098 on Build GRCh38
rs191369953 Chr.2: 97657994-97657994 on Build GRCh38
rs577318131 Chr.2: 97658005-97658005 on Build GRCh38
rs754195764 Chr.2: 97657990-97657990 on Build GRCh38
rs1016512288 Chr.2: 97658031-97658031 on Build GRCh38
rs757328064 Chr.2: 97658105-97658105 on Build GRCh38
rs757416072 Chr.2: 97658016-97658016 on Build GRCh38
rs766871606 Chr.2: 97657989-97657989 on Build GRCh38
rs140435232 Chr.2: 97658066-97658066 on Build GRCh38
rs1046962974 Chr.2: 97658025-97658025 on Build GRCh38
rs780973427 Chr.2: 97658021-97658021 on Build GRCh38
rs775404286 Chr.2: 97657973-97657973 on Build GRCh38
rs773384868 Chr.2: 97658100-97658102 on Build GRCh38
rs929905927 Chr.2: 97658020-97658020 on Build GRCh38
rs769762491 Chr.2: 97658018-97658020 on Build GRCh38
rs773684866 Chr.2: 97658061-97658061 on Build GRCh38
rs760033478 Chr.2: 97658067-97658067 on Build GRCh38
rs765672091 Chr.2: 97658073-97658073 on Build GRCh38
rs572322477 Chr.2: 97657971-97657971 on Build GRCh38
rs747173514 Chr.2: 97657954-97657954 on Build GRCh38
rs765908883 Chr.2: 97657996-97657996 on Build GRCh38
rs763325139 Chr.2: 97658090-97658090 on Build GRCh38
rs1040444916 Chr.2: 97658065-97658065 on Build GRCh38
rs754677033 Chr.2: 97658077-97658077 on Build GRCh38
rs770749832 Chr.2: 97657956-97657956 on Build GRCh38
rs745732438 Chr.2: 97658024-97658024 on Build GRCh38
rs755492665 Chr.2: 97657933-97657933 on Build GRCh38
rs200672180 Chr.2: 97657953-97657953 on Build GRCh38
rs755544675 Chr.2: 97657993-97657993 on Build GRCh38
rs762732953 Chr.2: 97657980-97657980 on Build GRCh38
rs144577602 Chr.2: 97658058-97658058 on Build GRCh38
rs1042705 Chr.2: 97658064-97658064 on Build GRCh38
rs781453544 Chr.2: 97658108-97658108 on Build GRCh38
rs758401310 Chr.2: 97657945-97657945 on Build GRCh38
rs761570777 Chr.2: 97658063-97658063 on Build GRCh38
rs959282737 Chr.2: 97658045-97658045 on Build GRCh38
rs924698234 Chr.2: 97658085-97658085 on Build GRCh38
rs747281452 Chr.2: 97658010-97658010 on Build GRCh38
rs748190438 Chr.2: 97657937-97657937 on Build GRCh38
rs543225703 Chr.2: 97658099-97658099 on Build GRCh38
rs149355636 Chr.2: 97657934-97657934 on Build GRCh38
rs772468833 Chr.2: 97657983-97657983 on Build GRCh38
rs753284851 Chr.2: 97658089-97658089 on Build GRCh38
rs768244227 Chr.2: 97658060-97658060 on Build GRCh38
rs780795599 Chr.2: 97658091-97658091 on Build GRCh38
rs775547948 Chr.2: 97658049-97658049 on Build GRCh38

View Full Product Details