Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 47242683-47242908 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACP2
Entrez Gene ID
53
Gene Name
acid phosphatase 2, lysosomal
Gene Aliases
LAP
UniGene
Hs.532492
Gene Chromosome Location
Chr.11: 47239302-47248847 on Build GRCh38
Transcripts
NM_001302489.1 NM_001302490.1 NM_001302491.1 NM_001302492.1 NM_001610.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5999712 c.983A>G p.Y328C Chr.11: 47242878-47242878 on Build GRCh38
1981313 c.1134C>T p.D378D Chr.11: 47242727-47242727 on Build GRCh38
6592429 c.1131A>G p.A377A Chr.11: 47242730-47242730 on Build GRCh38
1981317 c.988C>T p.R330W Chr.11: 47242873-47242873 on Build GRCh38
4033133 c.969C>T p.F323F Chr.11: 47242892-47242892 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1012901407 Chr.11: 47242718-47242718 on Build GRCh38
rs771791806 Chr.11: 47242876-47242876 on Build GRCh38
rs369759837 Chr.11: 47242788-47242788 on Build GRCh38
rs931544360 Chr.11: 47242865-47242865 on Build GRCh38
rs371800568 Chr.11: 47242710-47242710 on Build GRCh38
rs144006108 Chr.11: 47242845-47242845 on Build GRCh38
rs372169625 Chr.11: 47242794-47242794 on Build GRCh38
rs577187592 Chr.11: 47242778-47242778 on Build GRCh38
rs201581669 Chr.11: 47242709-47242709 on Build GRCh38
rs112139605 Chr.11: 47242766-47242766 on Build GRCh38
rs370922463 Chr.11: 47242738-47242738 on Build GRCh38
rs745556094 Chr.11: 47242786-47242786 on Build GRCh38
rs767798477 Chr.11: 47242706-47242706 on Build GRCh38
rs760594160 Chr.11: 47242716-47242716 on Build GRCh38
rs768231627 Chr.11: 47242885-47242885 on Build GRCh38
rs779487060 Chr.11: 47242872-47242872 on Build GRCh38
rs771454668 Chr.11: 47242780-47242780 on Build GRCh38
rs751798902 Chr.11: 47242737-47242737 on Build GRCh38
rs778103162 Chr.11: 47242866-47242866 on Build GRCh38
rs761469754 Chr.11: 47242733-47242733 on Build GRCh38
rs149746754 Chr.11: 47242739-47242739 on Build GRCh38
rs759077242 Chr.11: 47242704-47242704 on Build GRCh38
rs746460537 Chr.11: 47242884-47242884 on Build GRCh38
rs767574040 Chr.11: 47242853-47242853 on Build GRCh38
rs774098536 Chr.11: 47242781-47242781 on Build GRCh38
rs140392764 Chr.11: 47242835-47242835 on Build GRCh38
rs986721885 Chr.11: 47242816-47242816 on Build GRCh38
rs765062000 Chr.11: 47242819-47242819 on Build GRCh38
rs145123059 Chr.11: 47242719-47242719 on Build GRCh38
rs199783896 Chr.11: 47242856-47242856 on Build GRCh38
rs754392893 Chr.11: 47242867-47242867 on Build GRCh38
rs369127025 Chr.11: 47242818-47242818 on Build GRCh38
rs756639898 Chr.11: 47242777-47242777 on Build GRCh38
rs564407962 Chr.11: 47242725-47242725 on Build GRCh38
rs748687137 Chr.11: 47242759-47242759 on Build GRCh38
rs757664408 Chr.11: 47242868-47242868 on Build GRCh38
rs375066326 Chr.11: 47242809-47242809 on Build GRCh38
rs117138208 Chr.11: 47242802-47242802 on Build GRCh38
rs776202927 Chr.11: 47242715-47242715 on Build GRCh38
rs368717352 Chr.11: 47242795-47242795 on Build GRCh38
rs146415412 Chr.11: 47242854-47242854 on Build GRCh38
rs1013895546 Chr.11: 47242740-47242740 on Build GRCh38
rs745500880 Chr.11: 47242873-47242873 on Build GRCh38

View Full Product Details