Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 158909892-158910151 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C6orf99
Entrez Gene ID
100130967
Gene Name
chromosome 6 open reading frame 99
Gene Aliases
yR211F11.1
UniGene
Hs.32804
Gene Chromosome Location
Chr.6: 158869865-158918115 on Build GRCh38
Transcripts
NM_001195032.1 NM_001302839.1 NM_001302840.1 NM_001302841.1 XM_005266788.4 XM_011535374.2 XM_017010142.1 XM_017010143.1 XM_017010144.1 XM_017010145.1 XM_017010146.1 XM_017010147.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3023000 c.280C>T p.R94C Chr.6: 158910115-158910115 on Build GRCh38
3023004 c.296G>A p.R99H Chr.6: 158910131-158910131 on Build GRCh38
5906770 c.274C>T p.P92S Chr.6: 158910109-158910109 on Build GRCh38
4542362 c.547G>A p.E183K Chr.6: 158910151-158910151 on Build GRCh38
5996830 c.137G>C p.R46T Chr.6: 158909972-158909972 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200814870 Chr.6: 158909947-158909947 on Build GRCh38
rs1001733011 Chr.6: 158910020-158910020 on Build GRCh38
rs978690947 Chr.6: 158910098-158910098 on Build GRCh38
rs907336328 Chr.6: 158909931-158909931 on Build GRCh38
rs967705523 Chr.6: 158910085-158910085 on Build GRCh38
rs868623709 Chr.6: 158910126-158910126 on Build GRCh38
rs1034084488 Chr.6: 158910026-158910026 on Build GRCh38
rs912084789 Chr.6: 158909927-158909927 on Build GRCh38
rs747172813 Chr.6: 158910116-158910116 on Build GRCh38
rs1015801272 Chr.6: 158909951-158909951 on Build GRCh38
rs151266845 Chr.6: 158909949-158909949 on Build GRCh38
rs773228108 Chr.6: 158910077-158910077 on Build GRCh38
rs772351638 Chr.6: 158910079-158910079 on Build GRCh38
rs751923841 Chr.6: 158909971-158909971 on Build GRCh38
rs888114451 Chr.6: 158910006-158910006 on Build GRCh38
rs925171004 Chr.6: 158910108-158910108 on Build GRCh38
rs954063212 Chr.6: 158910033-158910033 on Build GRCh38
rs1028259736 Chr.6: 158909995-158909995 on Build GRCh38
rs547906908 Chr.6: 158910115-158910115 on Build GRCh38
rs754910693 Chr.6: 158909953-158909954 on Build GRCh38
rs747575954 Chr.6: 158910073-158910073 on Build GRCh38
rs978322649 Chr.6: 158910080-158910080 on Build GRCh38
rs753788705 Chr.6: 158909943-158909944 on Build GRCh38
rs963108452 Chr.6: 158909972-158909972 on Build GRCh38
rs144193169 Chr.6: 158910034-158910034 on Build GRCh38
rs759675957 Chr.6: 158909921-158909921 on Build GRCh38
rs778213087 Chr.6: 158909965-158909965 on Build GRCh38
rs12526317 Chr.6: 158909994-158909994 on Build GRCh38
rs199951771 Chr.6: 158910113-158910113 on Build GRCh38
rs1033466441 Chr.6: 158910112-158910112 on Build GRCh38
rs778531058 Chr.6: 158910044-158910044 on Build GRCh38
rs772145355 Chr.6: 158910076-158910076 on Build GRCh38
rs765326461 Chr.6: 158910130-158910130 on Build GRCh38
rs764798889 Chr.6: 158909954-158909954 on Build GRCh38
rs781520790 Chr.6: 158910019-158910019 on Build GRCh38
rs768158720 Chr.6: 158910041-158910041 on Build GRCh38
rs765398511 Chr.6: 158909938-158909938 on Build GRCh38
rs760679338 Chr.6: 158910095-158910095 on Build GRCh38
rs758965960 Chr.6: 158909953-158909953 on Build GRCh38

View Full Product Details