Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 179050168-179050439 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM20B
Entrez Gene ID
9917
Gene Name
family with sequence similarity 20 member B
Gene Aliases
gxk1
UniGene
Hs.5737
Gene Chromosome Location
Chr.1: 179025804-179076574 on Build GRCh38
Transcripts
NM_001324310.1 NM_001324311.1 NM_014864.3 XM_017003001.1 XM_017003002.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1473057 c.386G>A p.R129Q Chr.1: 179050287-179050287 on Build GRCh38
6659408 c.411G>A p.P137P Chr.1: 179050312-179050312 on Build GRCh38
7228719 c.378-3C>G p.? Chr.1: 179050276-179050276 on Build GRCh38
1601295 c.434A>G p.N145S Chr.1: 179050335-179050335 on Build GRCh38
6444851 c.384C>T p.S128S Chr.1: 179050285-179050285 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767626681 Chr.1: 179050339-179050339 on Build GRCh38
rs761805950 Chr.1: 179050293-179050293 on Build GRCh38
rs375570180 Chr.1: 179050370-179050370 on Build GRCh38
rs985566981 Chr.1: 179050196-179050196 on Build GRCh38
rs780931438 Chr.1: 179050389-179050389 on Build GRCh38
rs202188103 Chr.1: 179050325-179050325 on Build GRCh38
rs12755539 Chr.1: 179050384-179050384 on Build GRCh38
rs61731084 Chr.1: 179050312-179050312 on Build GRCh38
rs748878990 Chr.1: 179050415-179050415 on Build GRCh38
rs374729574 Chr.1: 179050249-179050249 on Build GRCh38
rs766480524 Chr.1: 179050355-179050355 on Build GRCh38
rs199839076 Chr.1: 179050215-179050215 on Build GRCh38
rs776494336 Chr.1: 179050283-179050283 on Build GRCh38
rs762829428 Chr.1: 179050306-179050306 on Build GRCh38
rs367951403 Chr.1: 179050200-179050200 on Build GRCh38
rs766037805 Chr.1: 179050394-179050394 on Build GRCh38
rs769440740 Chr.1: 179050292-179050292 on Build GRCh38
rs140637589 Chr.1: 179050231-179050231 on Build GRCh38
rs756250085 Chr.1: 179050247-179050247 on Build GRCh38
rs750467924 Chr.1: 179050243-179050243 on Build GRCh38
rs372697245 Chr.1: 179050395-179050395 on Build GRCh38
rs779514450 Chr.1: 179050369-179050369 on Build GRCh38
rs769437493 Chr.1: 179050343-179050343 on Build GRCh38
rs547559756 Chr.1: 179050368-179050368 on Build GRCh38
rs981113490 Chr.1: 179050357-179050357 on Build GRCh38
rs779172644 Chr.1: 179050266-179050266 on Build GRCh38
rs749803921 Chr.1: 179050256-179050256 on Build GRCh38
rs754298364 Chr.1: 179050366-179050366 on Build GRCh38
rs527886315 Chr.1: 179050203-179050203 on Build GRCh38
rs774403516 Chr.1: 179050201-179050201 on Build GRCh38
rs745360443 Chr.1: 179050287-179050287 on Build GRCh38
rs755333688 Chr.1: 179050263-179050263 on Build GRCh38
rs745519213 Chr.1: 179050390-179050390 on Build GRCh38
rs756123122 Chr.1: 179050351-179050351 on Build GRCh38
rs770460238 Chr.1: 179050282-179050282 on Build GRCh38
rs750700295 Chr.1: 179050346-179050346 on Build GRCh38
rs561208083 Chr.1: 179050195-179050195 on Build GRCh38
rs150031972 Chr.1: 179050311-179050311 on Build GRCh38
rs1020060175 Chr.1: 179050281-179050281 on Build GRCh38
rs781522382 Chr.1: 179050239-179050239 on Build GRCh38
rs757437703 Chr.1: 179050233-179050233 on Build GRCh38
rs750172461 Chr.1: 179050360-179050360 on Build GRCh38
rs1056842473 Chr.1: 179050294-179050294 on Build GRCh38
rs201763886 Chr.1: 179050276-179050276 on Build GRCh38

View Full Product Details