Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 3616891-3617126 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRACR2A
Entrez Gene ID
84766
Gene Name
calcium release activated channel regulator 2A
Gene Aliases
EFCAB4B
UniGene
Hs.504534
Gene Chromosome Location
Chr.12: 3613366-3753216 on Build GRCh38
Transcripts
NM_001144958.1 XM_006719021.3 XM_011521034.2 XM_011521036.2 XM_011521037.2 XM_011521038.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1562140 c.2065G>A p.A689T Chr.12: 3617000-3617000 on Build GRCh38
3981017 c.2089G>C p.E697Q Chr.12: 3616976-3616976 on Build GRCh38
5478213 c.2111+2T>C p.? Chr.12: 3616952-3616952 on Build GRCh38
7426543 c.2054A>T p.Y685F Chr.12: 3617011-3617011 on Build GRCh38
6339488 c.2085C>A p.T695T Chr.12: 3616980-3616980 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs773902524 Chr.12: 3617023-3617023 on Build GRCh38
rs886447751 Chr.12: 3617000-3617000 on Build GRCh38
rs35527523 Chr.12: 3616933-3616933 on Build GRCh38
rs753638230 Chr.12: 3617081-3617081 on Build GRCh38
rs1047758304 Chr.12: 3616960-3616960 on Build GRCh38
rs536450210 Chr.12: 3616923-3616923 on Build GRCh38
rs913538490 Chr.12: 3617065-3617065 on Build GRCh38
rs927899740 Chr.12: 3617089-3617089 on Build GRCh38
rs150985998 Chr.12: 3617001-3617001 on Build GRCh38
rs1047192917 Chr.12: 3617082-3617082 on Build GRCh38
rs759604444 Chr.12: 3616923-3616923 on Build GRCh38
rs182205274 Chr.12: 3616919-3616919 on Build GRCh38
rs1019225842 Chr.12: 3617016-3617016 on Build GRCh38
rs867842228 Chr.12: 3617075-3617075 on Build GRCh38
rs377405599 Chr.12: 3617051-3617051 on Build GRCh38
rs769646102 Chr.12: 3616966-3616966 on Build GRCh38
rs996734559 Chr.12: 3617034-3617034 on Build GRCh38
rs542795415 Chr.12: 3617020-3617020 on Build GRCh38
rs994954434 Chr.12: 3616944-3616944 on Build GRCh38
rs981973043 Chr.12: 3617084-3617084 on Build GRCh38
rs773120957 Chr.12: 3616995-3616995 on Build GRCh38
rs775218297 Chr.12: 3617078-3617078 on Build GRCh38
rs556115315 Chr.12: 3617064-3617064 on Build GRCh38
rs1033203596 Chr.12: 3617036-3617036 on Build GRCh38
rs891309954 Chr.12: 3617100-3617100 on Build GRCh38
rs369414947 Chr.12: 3617050-3617050 on Build GRCh38

View Full Product Details