Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67439675-67439936 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CORO1B
Entrez Gene ID
57175
Gene Name
coronin 1B
Gene Aliases
CORONIN-2
UniGene
Hs.6191
Gene Chromosome Location
Chr.11: 67438047-67443821 on Build GRCh38
Transcripts
NM_001018070.2 NM_020441.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6347327 c.1052C>A p.T351N Chr.11: 67439799-67439799 on Build GRCh38
5635312 c.1045G>A p.V349I Chr.11: 67439806-67439806 on Build GRCh38
1605113 c.1026G>T p.E342D Chr.11: 67439825-67439825 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs57866382 Chr.11: 67439726-67439726 on Build GRCh38
rs369560889 Chr.11: 67439764-67439764 on Build GRCh38
rs768673496 Chr.11: 67439702-67439702 on Build GRCh38
rs558234704 Chr.11: 67439767-67439767 on Build GRCh38
rs376691338 Chr.11: 67439718-67439718 on Build GRCh38
rs375419977 Chr.11: 67439739-67439739 on Build GRCh38
rs764700791 Chr.11: 67439884-67439884 on Build GRCh38
rs953926935 Chr.11: 67439728-67439728 on Build GRCh38
rs199982791 Chr.11: 67439806-67439806 on Build GRCh38
rs753391106 Chr.11: 67439840-67439840 on Build GRCh38
rs767050168 Chr.11: 67439810-67439810 on Build GRCh38
rs949652914 Chr.11: 67439777-67439777 on Build GRCh38
rs377479557 Chr.11: 67439882-67439882 on Build GRCh38
rs1015076884 Chr.11: 67439875-67439875 on Build GRCh38
rs113031617 Chr.11: 67439784-67439784 on Build GRCh38
rs202003575 Chr.11: 67439846-67439846 on Build GRCh38
rs780461731 Chr.11: 67439818-67439818 on Build GRCh38
rs199523951 Chr.11: 67439766-67439766 on Build GRCh38
rs976353070 Chr.11: 67439716-67439716 on Build GRCh38
rs372734741 Chr.11: 67439773-67439773 on Build GRCh38
rs143227306 Chr.11: 67439824-67439824 on Build GRCh38
rs562606407 Chr.11: 67439894-67439894 on Build GRCh38
rs756498675 Chr.11: 67439891-67439891 on Build GRCh38
rs965268730 Chr.11: 67439714-67439714 on Build GRCh38
rs760686943 Chr.11: 67439762-67439762 on Build GRCh38
rs759083840 Chr.11: 67439883-67439884 on Build GRCh38
rs368869832 Chr.11: 67439858-67439858 on Build GRCh38
rs763068547 Chr.11: 67439863-67439863 on Build GRCh38
rs144273420 Chr.11: 67439738-67439738 on Build GRCh38
rs760428587 Chr.11: 67439856-67439856 on Build GRCh38
rs751843564 Chr.11: 67439778-67439778 on Build GRCh38
rs776768134 Chr.11: 67439860-67439860 on Build GRCh38
rs773930796 Chr.11: 67439837-67439837 on Build GRCh38
rs368860428 Chr.11: 67439876-67439876 on Build GRCh38
rs771719209 Chr.11: 67439848-67439848 on Build GRCh38
rs541854415 Chr.11: 67439851-67439851 on Build GRCh38
rs757627508 Chr.11: 67439803-67439803 on Build GRCh38
rs111752638 Chr.11: 67439785-67439785 on Build GRCh38
rs372686577 Chr.11: 67439866-67439866 on Build GRCh38
rs748787820 Chr.11: 67439823-67439823 on Build GRCh38
rs557327122 Chr.11: 67439890-67439890 on Build GRCh38
rs112327198 Chr.11: 67439845-67439845 on Build GRCh38
rs757520831 Chr.11: 67439889-67439889 on Build GRCh38
rs749804039 Chr.11: 67439895-67439895 on Build GRCh38
rs746308979 Chr.11: 67439807-67439807 on Build GRCh38

View Full Product Details