Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 74364132-74364397 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDAP1
Entrez Gene ID
54332
Gene Name
ganglioside induced differentiation associated protein 1
Gene Aliases
CMT4, CMT4A, CMTRIA
UniGene
Hs.168950
Gene Chromosome Location
Chr.8: 74349251-74488867 on Build GRCh38
Transcripts
NM_001040875.2 NM_018972.2 NR_046346.1 XM_011517551.2 XM_011517552.2 XM_017013585.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5037082 c.1037G>T p.S346I Chr.8: 74364327-74364327 on Build GRCh38
5979543 c.1062A>T p.R354S Chr.8: 74364352-74364352 on Build GRCh38
5006352 c.928C>T p.R310W Chr.8: 74364218-74364218 on Build GRCh38
2875688 c.939G>A p.K313K Chr.8: 74364229-74364229 on Build GRCh38
1101596 c.991G>A p.G331R Chr.8: 74364281-74364281 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771751303 Chr.8: 74364232-74364232 on Build GRCh38
rs747927220 Chr.8: 74364288-74364288 on Build GRCh38
rs768729647 Chr.8: 74364256-74364256 on Build GRCh38
rs745516657 Chr.8: 74364205-74364205 on Build GRCh38
rs371960254 Chr.8: 74364254-74364254 on Build GRCh38
rs750616418 Chr.8: 74364195-74364196 on Build GRCh38
rs756599272 Chr.8: 74364184-74364184 on Build GRCh38
rs759748392 Chr.8: 74364304-74364304 on Build GRCh38
rs909730199 Chr.8: 74364266-74364266 on Build GRCh38
rs778448664 Chr.8: 74364202-74364202 on Build GRCh38
rs112786851 Chr.8: 74364329-74364329 on Build GRCh38
rs757607617 Chr.8: 74364352-74364352 on Build GRCh38
rs765609133 Chr.8: 74364356-74364356 on Build GRCh38
rs752758036 Chr.8: 74364175-74364175 on Build GRCh38
rs764613529 Chr.8: 74364340-74364340 on Build GRCh38
rs976792279 Chr.8: 74364170-74364170 on Build GRCh38
rs140811185 Chr.8: 74364296-74364296 on Build GRCh38
rs199529910 Chr.8: 74364255-74364255 on Build GRCh38
rs756461496 Chr.8: 74364305-74364306 on Build GRCh38
rs368943246 Chr.8: 74364335-74364335 on Build GRCh38
rs774476301 Chr.8: 74364298-74364298 on Build GRCh38
rs754319162 Chr.8: 74364349-74364349 on Build GRCh38
rs397515432 Chr.8: 74364270-74364270 on Build GRCh38
rs776565871 Chr.8: 74364263-74364263 on Build GRCh38
rs879254196 Chr.8: 74364275-74364275 on Build GRCh38
rs778192707 Chr.8: 74364187-74364187 on Build GRCh38
rs775559344 Chr.8: 74364332-74364332 on Build GRCh38
rs368055667 Chr.8: 74364197-74364197 on Build GRCh38
rs903539031 Chr.8: 74364199-74364199 on Build GRCh38
rs767553697 Chr.8: 74364164-74364164 on Build GRCh38
rs538389475 Chr.8: 74364218-74364218 on Build GRCh38
rs750114170 Chr.8: 74364371-74364371 on Build GRCh38
rs757597998 Chr.8: 74364200-74364200 on Build GRCh38

View Full Product Details