Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 80286270-80286537 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF5
Entrez Gene ID
2250
Gene Name
fibroblast growth factor 5
Gene Aliases
HBGF-5, Smag-82, TCMGLY
UniGene
Hs.37055
Gene Chromosome Location
Chr.4: 80266588-80291017 on Build GRCh38
Transcripts
NM_001291812.1 NM_004464.3 NM_033143.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4684534 c.523G>A p.A175T Chr.4: 80286388-80286388 on Build GRCh38
6663569 c.606C>T p.C202C Chr.4: 80286471-80286471 on Build GRCh38
1431303 c.544_545insA p.T184fs*11 Chr.4: 80286409-80286410 on Build GRCh38
6999879 c.518C>A p.T173N Chr.4: 80286383-80286383 on Build GRCh38
481569 c.493C>T p.R165C Chr.4: 80286358-80286358 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754259509 Chr.4: 80286307-80286307 on Build GRCh38
rs745662451 Chr.4: 80286359-80286359 on Build GRCh38
rs769814158 Chr.4: 80286371-80286371 on Build GRCh38
rs548602803 Chr.4: 80286358-80286358 on Build GRCh38
rs151173058 Chr.4: 80286516-80286516 on Build GRCh38
rs766726081 Chr.4: 80286299-80286299 on Build GRCh38
rs372468085 Chr.4: 80286378-80286378 on Build GRCh38
rs377522118 Chr.4: 80286294-80286294 on Build GRCh38
rs745607469 Chr.4: 80286449-80286449 on Build GRCh38
rs753110821 Chr.4: 80286421-80286421 on Build GRCh38
rs776917011 Chr.4: 80286415-80286415 on Build GRCh38
rs376649641 Chr.4: 80286492-80286492 on Build GRCh38
rs776672648 Chr.4: 80286409-80286410 on Build GRCh38
rs1030361837 Chr.4: 80286298-80286298 on Build GRCh38
rs781595517 Chr.4: 80286437-80286437 on Build GRCh38
rs1008034685 Chr.4: 80286329-80286329 on Build GRCh38
rs764152136 Chr.4: 80286306-80286307 on Build GRCh38
rs759008533 Chr.4: 80286422-80286422 on Build GRCh38
rs573054858 Chr.4: 80286475-80286475 on Build GRCh38
rs34120491 Chr.4: 80286319-80286319 on Build GRCh38
rs145032506 Chr.4: 80286388-80286388 on Build GRCh38
rs201742375 Chr.4: 80286402-80286402 on Build GRCh38
rs775311039 Chr.4: 80286377-80286377 on Build GRCh38
rs756921324 Chr.4: 80286408-80286408 on Build GRCh38
rs149079287 Chr.4: 80286435-80286435 on Build GRCh38
rs377127455 Chr.4: 80286416-80286416 on Build GRCh38
rs771675935 Chr.4: 80286474-80286474 on Build GRCh38
rs587777581 Chr.4: 80286385-80286385 on Build GRCh38
rs923946868 Chr.4: 80286452-80286452 on Build GRCh38
rs955234088 Chr.4: 80286308-80286308 on Build GRCh38
rs775977168 Chr.4: 80286478-80286478 on Build GRCh38
rs757607151 Chr.4: 80286357-80286357 on Build GRCh38
rs900532451 Chr.4: 80286496-80286496 on Build GRCh38
rs767752536 Chr.4: 80286517-80286517 on Build GRCh38
rs768021806 Chr.4: 80286387-80286387 on Build GRCh38
rs996564751 Chr.4: 80286500-80286500 on Build GRCh38
rs879554173 Chr.4: 80286504-80286504 on Build GRCh38
rs368204813 Chr.4: 80286341-80286341 on Build GRCh38
rs369250747 Chr.4: 80286425-80286425 on Build GRCh38
rs764750296 Chr.4: 80286515-80286525 on Build GRCh38
rs765511510 Chr.4: 80286477-80286477 on Build GRCh38
rs201557946 Chr.4: 80286464-80286464 on Build GRCh38
rs758383050 Chr.4: 80286317-80286317 on Build GRCh38
rs144149346 Chr.4: 80286348-80286348 on Build GRCh38
rs1038287777 Chr.4: 80286491-80286491 on Build GRCh38
rs752784451 Chr.4: 80286316-80286316 on Build GRCh38
rs200918179 Chr.4: 80286491-80286491 on Build GRCh38
rs747665760 Chr.4: 80286468-80286468 on Build GRCh38
rs200891724 Chr.4: 80286463-80286463 on Build GRCh38
rs765741235 Chr.4: 80286309-80286309 on Build GRCh38
rs376474142 Chr.4: 80286450-80286450 on Build GRCh38
rs761442219 Chr.4: 80286393-80286393 on Build GRCh38
rs767175395 Chr.4: 80286398-80286398 on Build GRCh38
rs201833676 Chr.4: 80286321-80286321 on Build GRCh38
rs373322040 Chr.4: 80286426-80286426 on Build GRCh38
rs761416300 Chr.4: 80286410-80286410 on Build GRCh38
rs928349231 Chr.4: 80286470-80286470 on Build GRCh38
rs955991250 Chr.4: 80286360-80286360 on Build GRCh38

View Full Product Details