Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 119229706-119229951 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B4GALT4
Entrez Gene ID
8702
Gene Name
beta-1,4-galactosyltransferase 4
Gene Aliases
B4Gal-T4, beta4Gal-T4
UniGene
Hs.13225
Gene Chromosome Location
Chr.3: 119211742-119240946 on Build GRCh38
Transcripts
NM_003778.3 NM_212543.1 XM_005247855.1 XM_006713798.2 XM_006713799.2 XM_006713800.1 XM_006713801.2 XM_011513260.1 XM_017007401.1 NR_046574.1

Gene Symbol
B4GALT4-AS1
Entrez Gene ID
100874201
Gene Name
B4GALT4 antisense RNA 1
Gene Aliases
-
UniGene
Hs.721307
Gene Chromosome Location
Chr.3: 119226486-119235141 on Build GRCh38
Transcripts
NM_003778.3 NM_212543.1 XM_005247855.1 XM_006713798.2 XM_006713799.2 XM_006713800.1 XM_006713801.2 XM_011513260.1 XM_017007401.1 NR_046574.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4112507 c.191C>T p.T64I Chr.3: 119229909-119229909 on Build GRCh38
4112506 c.206C>T p.T69M Chr.3: 119229894-119229894 on Build GRCh38
7147998 c.159G>A p.K53K Chr.3: 119229941-119229941 on Build GRCh38
4953257 c.187C>A p.L63M Chr.3: 119229913-119229913 on Build GRCh38
1168978 c.155A>G p.H52R Chr.3: 119229945-119229945 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370499485 Chr.3: 119229810-119229810 on Build GRCh38
rs865847574 Chr.3: 119229902-119229903 on Build GRCh38
rs928476883 Chr.3: 119229773-119229773 on Build GRCh38
rs758939498 Chr.3: 119229916-119229916 on Build GRCh38
rs778187476 Chr.3: 119229796-119229796 on Build GRCh38
rs4422294 Chr.3: 119229869-119229869 on Build GRCh38
rs776165671 Chr.3: 119229813-119229813 on Build GRCh38
rs910569200 Chr.3: 119229920-119229920 on Build GRCh38
rs752834059 Chr.3: 119229911-119229911 on Build GRCh38
rs775178147 Chr.3: 119229825-119229825 on Build GRCh38
rs778148279 Chr.3: 119229924-119229924 on Build GRCh38
rs760149633 Chr.3: 119229816-119229819 on Build GRCh38
rs745325529 Chr.3: 119229816-119229816 on Build GRCh38
rs755009310 Chr.3: 119229876-119229876 on Build GRCh38
rs961101053 Chr.3: 119229854-119229854 on Build GRCh38
rs933906771 Chr.3: 119229898-119229898 on Build GRCh38
rs545601169 Chr.3: 119229821-119229824 on Build GRCh38
rs765280694 Chr.3: 119229884-119229884 on Build GRCh38
rs753010844 Chr.3: 119229839-119229840 on Build GRCh38
rs935415667 Chr.3: 119229896-119229896 on Build GRCh38
rs761171544 Chr.3: 119229903-119229904 on Build GRCh38
rs769463342 Chr.3: 119229817-119229817 on Build GRCh38
rs779603446 Chr.3: 119229814-119229814 on Build GRCh38
rs770554706 Chr.3: 119229811-119229811 on Build GRCh38
rs754079803 Chr.3: 119229872-119229872 on Build GRCh38
rs762958936 Chr.3: 119229835-119229835 on Build GRCh38
rs149948131 Chr.3: 119229870-119229870 on Build GRCh38
rs755536437 Chr.3: 119229783-119229783 on Build GRCh38
rs761517196 Chr.3: 119229860-119229860 on Build GRCh38
rs201212531 Chr.3: 119229848-119229848 on Build GRCh38
rs747344665 Chr.3: 119229925-119229925 on Build GRCh38
rs4422293 Chr.3: 119229771-119229771 on Build GRCh38
rs763991177 Chr.3: 119229842-119229842 on Build GRCh38

View Full Product Details