Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 72206374-72206617 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERCC6L
Entrez Gene ID
54821
Gene Name
ERCC excision repair 6 like, spindle assembly checkpoint helicase
Gene Aliases
PICH, RAD26L
UniGene
Hs.47558
Gene Chromosome Location
Chr.X: 72204657-72239008 on Build GRCh38
Transcripts
NM_017669.2 NM_001170747.1

Gene Symbol
PIN4
Entrez Gene ID
5303
Gene Name
peptidylprolyl cis/trans isomerase, NIMA-interacting 4
Gene Aliases
EPVH, PAR14, PAR17
UniGene
Hs.655623
Gene Chromosome Location
Chr.X: 72181676-72263964 on Build GRCh38
Transcripts
NM_017669.2 NM_001170747.1

SNP Detail

SNP ID Chromosome Location
rs780115534 Chr.X: 72206433-72206433 on Build GRCh38
rs773634562 Chr.X: 72206481-72206481 on Build GRCh38
rs748332306 Chr.X: 72206462-72206462 on Build GRCh38
rs369492714 Chr.X: 72206509-72206509 on Build GRCh38
rs373087197 Chr.X: 72206426-72206426 on Build GRCh38
rs45448501 Chr.X: 72206491-72206491 on Build GRCh38
rs755042517 Chr.X: 72206439-72206439 on Build GRCh38
rs777094514 Chr.X: 72206518-72206518 on Build GRCh38
rs763537928 Chr.X: 72206584-72206584 on Build GRCh38
rs1026752035 Chr.X: 72206538-72206538 on Build GRCh38
rs375177556 Chr.X: 72206479-72206479 on Build GRCh38
rs779001189 Chr.X: 72206446-72206446 on Build GRCh38
rs138539407 Chr.X: 72206437-72206437 on Build GRCh38
rs760051206 Chr.X: 72206564-72206564 on Build GRCh38

View Full Product Details