Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 52024528-52024775 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALG11
Entrez Gene ID
440138
Gene Name
ALG11, alpha-1,2-mannosyltransferase
Gene Aliases
CDG1P, GT8
UniGene
Hs.512963
Gene Chromosome Location
Chr.13: 52012387-52029648 on Build GRCh38
Transcripts
NM_001004127.2 NR_036571.2 NM_021645.5

Gene Symbol
UTP14C
Entrez Gene ID
9724
Gene Name
UTP14, small subunit processome component homolog C (S. cerevisiae)
Gene Aliases
2700066J21Rik, KIAA0266, UTP14B
UniGene
Hs.512963
Gene Chromosome Location
Chr.13: 52024691-52033600 on Build GRCh38
Transcripts
NM_001004127.2 NR_036571.2 NM_021645.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
286417 c.926T>G p.F309C Chr.13: 52024656-52024656 on Build GRCh38
5822240 c.1042A>G p.K348E Chr.13: 52024772-52024772 on Build GRCh38
6537963 c.948A>G p.P316P Chr.13: 52024678-52024678 on Build GRCh38
1191067 c.896_897insC p.G300fs*11 Chr.13: 52024626-52024627 on Build GRCh38
1639368 c.974_976delTGC p.L326delL Chr.13: 52024704-52024706 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776577304 Chr.13: 52024561-52024561 on Build GRCh38
rs752964732 Chr.13: 52024591-52024591 on Build GRCh38
rs387907184 Chr.13: 52024683-52024683 on Build GRCh38
rs150850994 Chr.13: 52024624-52024624 on Build GRCh38
rs772433343 Chr.13: 52024646-52024646 on Build GRCh38
rs387907181 Chr.13: 52024566-52024566 on Build GRCh38
rs749941774 Chr.13: 52024618-52024618 on Build GRCh38
rs533408732 Chr.13: 52024604-52024604 on Build GRCh38
rs762701372 Chr.13: 52024592-52024592 on Build GRCh38
rs747485062 Chr.13: 52024648-52024648 on Build GRCh38
rs201547039 Chr.13: 52024574-52024574 on Build GRCh38
rs753485735 Chr.13: 52024750-52024750 on Build GRCh38
rs775647247 Chr.13: 52024680-52024680 on Build GRCh38
rs991088234 Chr.13: 52024715-52024715 on Build GRCh38
rs774018074 Chr.13: 52024707-52024707 on Build GRCh38
rs748980888 Chr.13: 52024637-52024637 on Build GRCh38
rs376118160 Chr.13: 52024630-52024630 on Build GRCh38
rs761729275 Chr.13: 52024710-52024710 on Build GRCh38
rs759676836 Chr.13: 52024562-52024562 on Build GRCh38
rs61958802 Chr.13: 52024663-52024663 on Build GRCh38
rs1021454171 Chr.13: 52024598-52024598 on Build GRCh38
rs763889797 Chr.13: 52024705-52024705 on Build GRCh38
rs778015185 Chr.13: 52024647-52024647 on Build GRCh38
rs540927215 Chr.13: 52024662-52024662 on Build GRCh38
rs769901623 Chr.13: 52024679-52024679 on Build GRCh38
rs759590601 Chr.13: 52024670-52024670 on Build GRCh38
rs373146305 Chr.13: 52024594-52024594 on Build GRCh38
rs765431074 Chr.13: 52024676-52024676 on Build GRCh38
rs921029114 Chr.13: 52024717-52024717 on Build GRCh38
rs757152270 Chr.13: 52024608-52024608 on Build GRCh38
rs750430228 Chr.13: 52024721-52024721 on Build GRCh38
rs763180583 Chr.13: 52024684-52024684 on Build GRCh38
rs767338407 Chr.13: 52024719-52024719 on Build GRCh38
rs766009227 Chr.13: 52024740-52024740 on Build GRCh38
rs1051308050 Chr.13: 52024730-52024730 on Build GRCh38
rs145540579 Chr.13: 52024737-52024737 on Build GRCh38
rs767720592 Chr.13: 52024613-52024613 on Build GRCh38
rs763823178 Chr.13: 52024600-52024600 on Build GRCh38
rs779824996 Chr.13: 52024636-52024636 on Build GRCh38
rs891276611 Chr.13: 52024738-52024738 on Build GRCh38
rs765432044 Chr.13: 52024582-52024582 on Build GRCh38
rs959802397 Chr.13: 52024689-52024689 on Build GRCh38

View Full Product Details