Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 36136148-36136391 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELP2
Entrez Gene ID
55250
Gene Name
elongator acetyltransferase complex subunit 2
Gene Aliases
SHINC-2, STATIP1, StIP
UniGene
Hs.606040
Gene Chromosome Location
Chr.18: 36129874-36177931 on Build GRCh38
Transcripts
NM_001242875.2 NM_001242876.2 NM_001242877.2 NM_001242878.2 NM_001242879.2 NM_001324465.1 NM_001324466.1 NM_001324467.1 NM_001324468.1 NM_018255.3 NR_040110.2 NR_136897.1 NR_136898.1 NR_137173.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
296288 c.222T>C p.P74P Chr.18: 36136311-36136311 on Build GRCh38
5833504 c.243delA p.G82fs*6 Chr.18: 36136332-36136332 on Build GRCh38
5992042 c.259delG p.V87fs*1 Chr.18: 36136348-36136348 on Build GRCh38
4453107 c.253A>T p.N85Y Chr.18: 36136342-36136342 on Build GRCh38
4453106 c.253A>T p.N85Y Chr.18: 36136342-36136342 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs976209591 Chr.18: 36136216-36136216 on Build GRCh38
rs768965169 Chr.18: 36136351-36136351 on Build GRCh38
rs779250000 Chr.18: 36136364-36136364 on Build GRCh38
rs754798447 Chr.18: 36136308-36136308 on Build GRCh38
rs201181836 Chr.18: 36136271-36136271 on Build GRCh38
rs910582352 Chr.18: 36136201-36136201 on Build GRCh38
rs781276353 Chr.18: 36136323-36136323 on Build GRCh38
rs528771713 Chr.18: 36136189-36136189 on Build GRCh38
rs752641127 Chr.18: 36136315-36136315 on Build GRCh38
rs749565006 Chr.18: 36136347-36136347 on Build GRCh38
rs756287485 Chr.18: 36136332-36136332 on Build GRCh38
rs753582203 Chr.18: 36136303-36136303 on Build GRCh38
rs924410340 Chr.18: 36136237-36136237 on Build GRCh38
rs762574072 Chr.18: 36136342-36136345 on Build GRCh38
rs750459760 Chr.18: 36136324-36136324 on Build GRCh38
rs760286494 Chr.18: 36136294-36136294 on Build GRCh38
rs531087532 Chr.18: 36136274-36136274 on Build GRCh38
rs200390715 Chr.18: 36136293-36136293 on Build GRCh38
rs766124505 Chr.18: 36136297-36136297 on Build GRCh38
rs986941974 Chr.18: 36136247-36136247 on Build GRCh38
rs780222162 Chr.18: 36136335-36136335 on Build GRCh38
rs762255223 Chr.18: 36136275-36136275 on Build GRCh38
rs775816583 Chr.18: 36136241-36136241 on Build GRCh38
rs374737826 Chr.18: 36136284-36136284 on Build GRCh38
rs775042386 Chr.18: 36136304-36136304 on Build GRCh38
rs758275789 Chr.18: 36136318-36136318 on Build GRCh38
rs74438152 Chr.18: 36136313-36136313 on Build GRCh38
rs777552776 Chr.18: 36136252-36136253 on Build GRCh38
rs766982643 Chr.18: 36136281-36136281 on Build GRCh38
rs138438795 Chr.18: 36136213-36136213 on Build GRCh38
rs749652917 Chr.18: 36136259-36136259 on Build GRCh38
rs549365521 Chr.18: 36136252-36136252 on Build GRCh38
rs999659208 Chr.18: 36136178-36136178 on Build GRCh38

View Full Product Details