Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 51354128-51354355 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ETFB
Entrez Gene ID
2109
Gene Name
electron transfer flavoprotein beta subunit
Gene Aliases
FP585, MADD
UniGene
Hs.348531
Gene Chromosome Location
Chr.19: 51345155-51366418 on Build GRCh38
Transcripts
NM_001014763.1 NM_001985.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
713193 c.335G>T p.R112L Chr.19: 51354304-51354304 on Build GRCh38
3743087 c.168G>T p.K56N Chr.19: 51354198-51354198 on Build GRCh38
349647 c.396C>G p.F132L Chr.19: 51354243-51354243 on Build GRCh38
3797435 c.192C>T p.V64V Chr.19: 51354174-51354174 on Build GRCh38
5667549 c.399T>C p.C133C Chr.19: 51354240-51354240 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs989233154 Chr.19: 51354176-51354176 on Build GRCh38
rs374935848 Chr.19: 51354234-51354234 on Build GRCh38
rs770467292 Chr.19: 51354164-51354164 on Build GRCh38
rs368212620 Chr.19: 51354207-51354207 on Build GRCh38
rs760573811 Chr.19: 51354197-51354197 on Build GRCh38
rs750230877 Chr.19: 51354284-51354284 on Build GRCh38
rs781748603 Chr.19: 51354149-51354149 on Build GRCh38
rs564020088 Chr.19: 51354288-51354288 on Build GRCh38
rs267605609 Chr.19: 51354177-51354177 on Build GRCh38
rs754545072 Chr.19: 51354302-51354302 on Build GRCh38
rs374819445 Chr.19: 51354285-51354285 on Build GRCh38
rs759494324 Chr.19: 51354183-51354183 on Build GRCh38
rs764083482 Chr.19: 51354198-51354198 on Build GRCh38
rs151108898 Chr.19: 51354233-51354233 on Build GRCh38
rs774387920 Chr.19: 51354242-51354242 on Build GRCh38
rs537142189 Chr.19: 51354228-51354228 on Build GRCh38
rs767608788 Chr.19: 51354192-51354192 on Build GRCh38
rs775702541 Chr.19: 51354334-51354334 on Build GRCh38
rs142966954 Chr.19: 51354179-51354179 on Build GRCh38
rs199860339 Chr.19: 51354306-51354306 on Build GRCh38
rs770567402 Chr.19: 51354185-51354187 on Build GRCh38
rs371751519 Chr.19: 51354274-51354274 on Build GRCh38
rs765099692 Chr.19: 51354281-51354281 on Build GRCh38
rs746082442 Chr.19: 51354244-51354244 on Build GRCh38
rs772322071 Chr.19: 51354248-51354248 on Build GRCh38
rs750117869 Chr.19: 51354224-51354224 on Build GRCh38
rs183159891 Chr.19: 51354273-51354273 on Build GRCh38
rs775541180 Chr.19: 51354253-51354253 on Build GRCh38
rs368139326 Chr.19: 51354268-51354268 on Build GRCh38
rs374288379 Chr.19: 51354305-51354305 on Build GRCh38
rs1005896775 Chr.19: 51354291-51354291 on Build GRCh38
rs553721514 Chr.19: 51354335-51354335 on Build GRCh38
rs746284138 Chr.19: 51354307-51354307 on Build GRCh38
rs752825481 Chr.19: 51354196-51354196 on Build GRCh38
rs374583902 Chr.19: 51354180-51354180 on Build GRCh38
rs745870211 Chr.19: 51354335-51354336 on Build GRCh38
rs760800809 Chr.19: 51354254-51354254 on Build GRCh38
rs748684435 Chr.19: 51354159-51354159 on Build GRCh38
rs371201213 Chr.19: 51354297-51354297 on Build GRCh38
rs369216610 Chr.19: 51354304-51354304 on Build GRCh38
rs796051954 Chr.19: 51354206-51354206 on Build GRCh38
rs757088247 Chr.19: 51354200-51354200 on Build GRCh38
rs199705168 Chr.19: 51354182-51354182 on Build GRCh38
rs770414295 Chr.19: 51354232-51354232 on Build GRCh38
rs372040033 Chr.19: 51354231-51354231 on Build GRCh38
rs780806600 Chr.19: 51354198-51354200 on Build GRCh38
rs772266930 Chr.19: 51354330-51354330 on Build GRCh38

View Full Product Details