Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 20090135-20090366 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGCR8
Entrez Gene ID
54487
Gene Name
DGCR8 microprocessor complex subunit
Gene Aliases
C22orf12, DGCRK6, Gy1, pasha
UniGene
Hs.643452
Gene Chromosome Location
Chr.22: 20080232-20111877 on Build GRCh38
Transcripts
NM_001190326.1 NM_022720.6 XM_006724268.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5486938 c.1238C>G p.A413G Chr.22: 20090190-20090190 on Build GRCh38
1032179 c.1259T>C p.L420P Chr.22: 20090211-20090211 on Build GRCh38
3732654 c.1282G>A p.E428K Chr.22: 20090234-20090234 on Build GRCh38
6642883 c.1216G>A p.D406N Chr.22: 20090168-20090168 on Build GRCh38
5440930 c.1240G>C p.E414Q Chr.22: 20090192-20090192 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764017771 Chr.22: 20090200-20090200 on Build GRCh38
rs765059553 Chr.22: 20090319-20090319 on Build GRCh38
rs754774134 Chr.22: 20090239-20090239 on Build GRCh38
rs775843667 Chr.22: 20090303-20090303 on Build GRCh38
rs190748841 Chr.22: 20090158-20090158 on Build GRCh38
rs746210066 Chr.22: 20090186-20090186 on Build GRCh38
rs200010909 Chr.22: 20090171-20090171 on Build GRCh38
rs967992945 Chr.22: 20090210-20090210 on Build GRCh38
rs536682730 Chr.22: 20090293-20090293 on Build GRCh38
rs113891618 Chr.22: 20090233-20090233 on Build GRCh38
rs777343509 Chr.22: 20090169-20090169 on Build GRCh38
rs771119825 Chr.22: 20090191-20090191 on Build GRCh38
rs1020588406 Chr.22: 20090334-20090334 on Build GRCh38
rs749708126 Chr.22: 20090307-20090307 on Build GRCh38
rs752333362 Chr.22: 20090248-20090248 on Build GRCh38
rs370744836 Chr.22: 20090290-20090290 on Build GRCh38
rs756087746 Chr.22: 20090157-20090157 on Build GRCh38
rs376929969 Chr.22: 20090183-20090183 on Build GRCh38
rs754864266 Chr.22: 20090162-20090162 on Build GRCh38
rs34204101 Chr.22: 20090197-20090197 on Build GRCh38
rs147234686 Chr.22: 20090243-20090243 on Build GRCh38
rs770260935 Chr.22: 20090212-20090212 on Build GRCh38
rs753685267 Chr.22: 20090160-20090160 on Build GRCh38
rs747416922 Chr.22: 20090170-20090170 on Build GRCh38
rs557013793 Chr.22: 20090333-20090333 on Build GRCh38
rs770165245 Chr.22: 20090189-20090189 on Build GRCh38
rs774472650 Chr.22: 20090207-20090207 on Build GRCh38
rs2073779 Chr.22: 20090340-20090340 on Build GRCh38
rs140543646 Chr.22: 20090164-20090164 on Build GRCh38
rs750259282 Chr.22: 20090156-20090156 on Build GRCh38
rs560910865 Chr.22: 20090166-20090166 on Build GRCh38
rs150434219 Chr.22: 20090167-20090167 on Build GRCh38
rs746495980 Chr.22: 20090276-20090276 on Build GRCh38
rs758082594 Chr.22: 20090254-20090254 on Build GRCh38
rs761868924 Chr.22: 20090214-20090214 on Build GRCh38
rs749300283 Chr.22: 20090181-20090182 on Build GRCh38
rs766313804 Chr.22: 20090224-20090224 on Build GRCh38
rs903137379 Chr.22: 20090335-20090335 on Build GRCh38
rs958822880 Chr.22: 20090194-20090194 on Build GRCh38
rs777348779 Chr.22: 20090255-20090255 on Build GRCh38
rs373684505 Chr.22: 20090302-20090302 on Build GRCh38

View Full Product Details