Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 15296633-15296898 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FHAD1
Entrez Gene ID
114827
Gene Name
forkhead associated phosphopeptide binding domain 1
Gene Aliases
-
UniGene
Hs.659997
Gene Chromosome Location
Chr.1: 15236559-15400283 on Build GRCh38
Transcripts
NM_052929.1 XM_011540575.2 XM_011540576.2 XM_011540577.2 XM_011540580.2 XM_011540581.2 XM_011540582.2 XM_011540584.2 XM_011540588.2 XM_011540589.2 XM_011540590.2 XM_011540592.2 XM_011540593.2 XM_011540595.2 XM_011540597.2 XM_011540598.2 XM_017000198.1 XM_017000199.1 XM_017000200.1 XM_017000201.1 XM_017000202.1 XM_017000203.1 XM_017000204.1 XM_017000205.1 XM_017000206.1 XM_017000207.1 XM_017000208.1 XM_017000209.1 XM_017000210.1 XM_017000211.1 XM_017000212.1 XM_017000213.1 XM_017000214.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6506945 c.70G>A p.E24K Chr.1: 15296767-15296767 on Build GRCh38
6506943 c.652G>A p.E218K Chr.1: 15296767-15296767 on Build GRCh38
3705075 c.614G>A p.G205E Chr.1: 15296729-15296729 on Build GRCh38
4802648 c.675C>G p.D225E Chr.1: 15296790-15296790 on Build GRCh38
4555371 c.69G>T p.V23V Chr.1: 15296766-15296766 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776442613 Chr.1: 15296779-15296779 on Build GRCh38
rs537984407 Chr.1: 15296787-15296787 on Build GRCh38
rs756440993 Chr.1: 15296706-15296706 on Build GRCh38
rs985686028 Chr.1: 15296748-15296748 on Build GRCh38
rs376640150 Chr.1: 15296725-15296725 on Build GRCh38
rs969071084 Chr.1: 15296747-15296747 on Build GRCh38
rs745474139 Chr.1: 15296782-15296782 on Build GRCh38
rs56925120 Chr.1: 15296840-15296844 on Build GRCh38
rs181060581 Chr.1: 15296878-15296878 on Build GRCh38
rs941629123 Chr.1: 15296857-15296857 on Build GRCh38
rs527694107 Chr.1: 15296717-15296717 on Build GRCh38
rs200196291 Chr.1: 15296772-15296772 on Build GRCh38
rs1045125127 Chr.1: 15296867-15296867 on Build GRCh38
rs775071554 Chr.1: 15296795-15296795 on Build GRCh38
rs768062365 Chr.1: 15296683-15296683 on Build GRCh38
rs762509836 Chr.1: 15296835-15296835 on Build GRCh38
rs564985288 Chr.1: 15296703-15296703 on Build GRCh38
rs948766923 Chr.1: 15296837-15296837 on Build GRCh38
rs199916877 Chr.1: 15296728-15296728 on Build GRCh38
rs968663603 Chr.1: 15296746-15296746 on Build GRCh38
rs769794956 Chr.1: 15296845-15296845 on Build GRCh38
rs746936223 Chr.1: 15296754-15296754 on Build GRCh38
rs779330850 Chr.1: 15296727-15296727 on Build GRCh38
rs35516622 Chr.1: 15296859-15296859 on Build GRCh38
rs929734844 Chr.1: 15296836-15296836 on Build GRCh38
rs12032022 Chr.1: 15296718-15296718 on Build GRCh38
rs79751240 Chr.1: 15296856-15296856 on Build GRCh38
rs529140966 Chr.1: 15296744-15296744 on Build GRCh38
rs549836891 Chr.1: 15296682-15296682 on Build GRCh38
rs762022768 Chr.1: 15296806-15296806 on Build GRCh38
rs978600894 Chr.1: 15296749-15296749 on Build GRCh38
rs549231708 Chr.1: 15296753-15296753 on Build GRCh38
rs145866454 Chr.1: 15296837-15296841 on Build GRCh38

View Full Product Details