Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41715134-41715369 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEACAM5
Entrez Gene ID
1048
Gene Name
carcinoembryonic antigen related cell adhesion molecule 5
Gene Aliases
CD66e, CEA
UniGene
Hs.709196
Gene Chromosome Location
Chr.19: 41708585-41730433 on Build GRCh38
Transcripts
NM_001291484.2 NM_001308398.1 NM_004363.5 XM_005258413.3 XM_011526322.1 XM_017026145.1 XM_017026146.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5921401 c.652C>T p.Q218* Chr.19: 41715198-41715198 on Build GRCh38
5501963 c.635G>A p.S212N Chr.19: 41715181-41715181 on Build GRCh38
4581088 c.672G>A p.R224R Chr.19: 41715218-41715218 on Build GRCh38
344707 c.632C>A p.A211E Chr.19: 41715178-41715178 on Build GRCh38
279799 c.673C>T p.R225C Chr.19: 41715219-41715219 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs960514834 Chr.19: 41715233-41715233 on Build GRCh38
rs571502155 Chr.19: 41715301-41715301 on Build GRCh38
rs527722854 Chr.19: 41715203-41715203 on Build GRCh38
rs372254328 Chr.19: 41715210-41715210 on Build GRCh38
rs17853315 Chr.19: 41715174-41715174 on Build GRCh38
rs782198510 Chr.19: 41715163-41715163 on Build GRCh38
rs782541248 Chr.19: 41715215-41715215 on Build GRCh38
rs782292675 Chr.19: 41715214-41715214 on Build GRCh38
rs782039954 Chr.19: 41715178-41715178 on Build GRCh38
rs375538670 Chr.19: 41715194-41715194 on Build GRCh38
rs369164921 Chr.19: 41715289-41715289 on Build GRCh38
rs782583835 Chr.19: 41715213-41715213 on Build GRCh38
rs781931742 Chr.19: 41715199-41715199 on Build GRCh38
rs782304728 Chr.19: 41715221-41715221 on Build GRCh38
rs782283892 Chr.19: 41715332-41715332 on Build GRCh38
rs782500077 Chr.19: 41715170-41715170 on Build GRCh38
rs782753397 Chr.19: 41715205-41715205 on Build GRCh38
rs781991272 Chr.19: 41715183-41715183 on Build GRCh38
rs781830094 Chr.19: 41715207-41715207 on Build GRCh38
rs782086805 Chr.19: 41715291-41715291 on Build GRCh38
rs184827523 Chr.19: 41715330-41715330 on Build GRCh38
rs1005474927 Chr.19: 41715181-41715181 on Build GRCh38
rs782453218 Chr.19: 41715329-41715329 on Build GRCh38
rs782403641 Chr.19: 41715348-41715348 on Build GRCh38
rs782718819 Chr.19: 41715267-41715267 on Build GRCh38
rs199564277 Chr.19: 41715238-41715238 on Build GRCh38
rs782788457 Chr.19: 41715300-41715300 on Build GRCh38
rs782093327 Chr.19: 41715189-41715189 on Build GRCh38
rs376770900 Chr.19: 41715220-41715220 on Build GRCh38
rs781821119 Chr.19: 41715318-41715318 on Build GRCh38
rs782511943 Chr.19: 41715204-41715204 on Build GRCh38
rs369870451 Chr.19: 41715201-41715201 on Build GRCh38
rs782136127 Chr.19: 41715232-41715232 on Build GRCh38
rs537615653 Chr.19: 41715273-41715273 on Build GRCh38
rs562617012 Chr.19: 41715166-41715166 on Build GRCh38
rs141597462 Chr.19: 41715202-41715202 on Build GRCh38
rs368912460 Chr.19: 41715236-41715236 on Build GRCh38
rs533268241 Chr.19: 41715180-41715180 on Build GRCh38
rs782602037 Chr.19: 41715160-41715160 on Build GRCh38
rs373238370 Chr.19: 41715255-41715255 on Build GRCh38
rs538715376 Chr.19: 41715306-41715306 on Build GRCh38
rs781976338 Chr.19: 41715242-41715242 on Build GRCh38
rs782508644 Chr.19: 41715302-41715302 on Build GRCh38

View Full Product Details