Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 34967126-34967397 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKS1A
Entrez Gene ID
23294
Gene Name
ankyrin repeat and sterile alpha motif domain containing 1A
Gene Aliases
ANKS1
UniGene
Hs.656492
Gene Chromosome Location
Chr.6: 34889261-35097994 on Build GRCh38
Transcripts
NM_015245.2 XM_005248964.3 XM_006715036.2 XM_011514431.2 XM_011514432.2 XM_011514433.1 XM_011514434.2 XM_011514435.2 XM_011514436.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3873389 c.247C>T p.P83S Chr.6: 34967288-34967288 on Build GRCh38
6471142 c.198-5delG p.? Chr.6: 34967234-34967234 on Build GRCh38
3735709 c.222C>A p.N74K Chr.6: 34967263-34967263 on Build GRCh38
1443959 c.273C>T p.G91G Chr.6: 34967314-34967314 on Build GRCh38
3411026 c.210G>A p.G70G Chr.6: 34967251-34967251 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs771209941 Chr.6: 34967211-34967211 on Build GRCh38
rs557544500 Chr.6: 34967342-34967342 on Build GRCh38
rs1032610789 Chr.6: 34967331-34967331 on Build GRCh38
rs756086879 Chr.6: 34967197-34967197 on Build GRCh38
rs748336813 Chr.6: 34967212-34967215 on Build GRCh38
rs769436196 Chr.6: 34967330-34967330 on Build GRCh38
rs769004352 Chr.6: 34967237-34967237 on Build GRCh38
rs900306642 Chr.6: 34967345-34967345 on Build GRCh38
rs776028064 Chr.6: 34967228-34967228 on Build GRCh38
rs754737714 Chr.6: 34967205-34967205 on Build GRCh38
rs371988677 Chr.6: 34967217-34967217 on Build GRCh38
rs375096952 Chr.6: 34967288-34967288 on Build GRCh38
rs760181911 Chr.6: 34967302-34967302 on Build GRCh38
rs781346976 Chr.6: 34967362-34967362 on Build GRCh38
rs552303940 Chr.6: 34967179-34967179 on Build GRCh38
rs147758055 Chr.6: 34967290-34967290 on Build GRCh38
rs757452730 Chr.6: 34967354-34967354 on Build GRCh38
rs536063772 Chr.6: 34967293-34967293 on Build GRCh38
rs1030229479 Chr.6: 34967353-34967353 on Build GRCh38
rs753753053 Chr.6: 34967329-34967329 on Build GRCh38
rs147203909 Chr.6: 34967181-34967181 on Build GRCh38
rs763372061 Chr.6: 34967229-34967229 on Build GRCh38
rs746465121 Chr.6: 34967233-34967233 on Build GRCh38
rs765814131 Chr.6: 34967314-34967314 on Build GRCh38
rs773361442 Chr.6: 34967218-34967221 on Build GRCh38
rs780087850 Chr.6: 34967200-34967200 on Build GRCh38
rs777360613 Chr.6: 34967219-34967220 on Build GRCh38
rs774910506 Chr.6: 34967239-34967239 on Build GRCh38
rs80085216 Chr.6: 34967195-34967195 on Build GRCh38
rs1042065166 Chr.6: 34967236-34967236 on Build GRCh38
rs753739794 Chr.6: 34967201-34967201 on Build GRCh38
rs758057996 Chr.6: 34967220-34967220 on Build GRCh38
rs781221575 Chr.6: 34967214-34967214 on Build GRCh38
rs80037942 Chr.6: 34967220-34967220 on Build GRCh38
rs778587722 Chr.6: 34967206-34967206 on Build GRCh38
rs761552683 Chr.6: 34967251-34967251 on Build GRCh38
rs370066201 Chr.6: 34967167-34967167 on Build GRCh38
rs777046450 Chr.6: 34967196-34967196 on Build GRCh38
rs367910044 Chr.6: 34967187-34967187 on Build GRCh38
rs145339705 Chr.6: 34967260-34967260 on Build GRCh38
rs745999447 Chr.6: 34967215-34967215 on Build GRCh38
rs184209306 Chr.6: 34967207-34967207 on Build GRCh38
rs138802691 Chr.6: 34967346-34967346 on Build GRCh38
rs761566982 Chr.6: 34967194-34967194 on Build GRCh38
rs769788967 Chr.6: 34967222-34967222 on Build GRCh38

View Full Product Details