Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 34345907-34346180 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUTM1
Entrez Gene ID
256646
Gene Name
NUT midline carcinoma family member 1
Gene Aliases
C15orf55, FAM22H, NUT
UniGene
Hs.525769
Gene Chromosome Location
Chr.15: 34343315-34357737 on Build GRCh38
Transcripts
NM_001284292.1 NM_001284293.1 NM_175741.2

SNP Detail

SNP ID Chromosome Location
rs773972237 Chr.15: 34345955-34345955 on Build GRCh38
rs759178769 Chr.15: 34345957-34345957 on Build GRCh38
rs767186692 Chr.15: 34345960-34345960 on Build GRCh38
rs925598806 Chr.15: 34346053-34346055 on Build GRCh38
rs762364000 Chr.15: 34345932-34345932 on Build GRCh38
rs12592913 Chr.15: 34346141-34346141 on Build GRCh38
rs781676149 Chr.15: 34346074-34346074 on Build GRCh38
rs762673130 Chr.15: 34346036-34346036 on Build GRCh38
rs780325233 Chr.15: 34346045-34346045 on Build GRCh38
rs763825935 Chr.15: 34345989-34345989 on Build GRCh38
rs376561921 Chr.15: 34346048-34346048 on Build GRCh38
rs753820990 Chr.15: 34345993-34345993 on Build GRCh38
rs748709103 Chr.15: 34346077-34346077 on Build GRCh38
rs758349512 Chr.15: 34346043-34346043 on Build GRCh38
rs368680128 Chr.15: 34345988-34345988 on Build GRCh38
rs1047125393 Chr.15: 34345962-34345962 on Build GRCh38
rs775073763 Chr.15: 34346010-34346010 on Build GRCh38
rs563275490 Chr.15: 34346134-34346134 on Build GRCh38
rs913157253 Chr.15: 34345931-34345931 on Build GRCh38
rs774949005 Chr.15: 34345961-34345961 on Build GRCh38
rs200279336 Chr.15: 34346035-34346035 on Build GRCh38
rs770618494 Chr.15: 34345942-34345942 on Build GRCh38
rs765368453 Chr.15: 34346019-34346019 on Build GRCh38
rs533257672 Chr.15: 34346106-34346106 on Build GRCh38
rs942468610 Chr.15: 34346110-34346110 on Build GRCh38
rs544999559 Chr.15: 34346131-34346131 on Build GRCh38
rs770420468 Chr.15: 34346084-34346084 on Build GRCh38
rs12595504 Chr.15: 34345997-34345997 on Build GRCh38
rs755419089 Chr.15: 34346064-34346064 on Build GRCh38
rs930180121 Chr.15: 34345933-34345933 on Build GRCh38

View Full Product Details