Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 82726782-82727009 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FN3KRP
Entrez Gene ID
79672
Gene Name
fructosamine 3 kinase related protein
Gene Aliases
FN3KL
UniGene
Hs.31431
Gene Chromosome Location
Chr.17: 82716706-82728017 on Build GRCh38
Transcripts
NM_024619.3 NR_046408.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5833476 c.702delC p.P235fs*14 Chr.17: 82726943-82726943 on Build GRCh38
5504399 c.706G>A p.A236T Chr.17: 82726947-82726947 on Build GRCh38
6888997 c.612C>T p.F204F Chr.17: 82726853-82726853 on Build GRCh38
2807083 c.745G>T p.A249S Chr.17: 82726986-82726986 on Build GRCh38
5727205 c.597G>A p.K199K Chr.17: 82726838-82726838 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748745004 Chr.17: 82726842-82726842 on Build GRCh38
rs370444990 Chr.17: 82726931-82726931 on Build GRCh38
rs773076464 Chr.17: 82726860-82726860 on Build GRCh38
rs774023407 Chr.17: 82726848-82726848 on Build GRCh38
rs35865076 Chr.17: 82726861-82726862 on Build GRCh38
rs768222352 Chr.17: 82726845-82726845 on Build GRCh38
rs138064117 Chr.17: 82726929-82726929 on Build GRCh38
rs749549834 Chr.17: 82726853-82726853 on Build GRCh38
rs1031813781 Chr.17: 82726971-82726971 on Build GRCh38
rs1006094295 Chr.17: 82726825-82726825 on Build GRCh38
rs146293327 Chr.17: 82726967-82726967 on Build GRCh38
rs765429684 Chr.17: 82726893-82726893 on Build GRCh38
rs138910887 Chr.17: 82726854-82726854 on Build GRCh38
rs371279348 Chr.17: 82726841-82726841 on Build GRCh38
rs556222572 Chr.17: 82726930-82726930 on Build GRCh38
rs771035632 Chr.17: 82726966-82726966 on Build GRCh38
rs746763211 Chr.17: 82726960-82726960 on Build GRCh38
rs202184366 Chr.17: 82726830-82726830 on Build GRCh38
rs771196672 Chr.17: 82726882-82726882 on Build GRCh38
rs939258008 Chr.17: 82726885-82726885 on Build GRCh38
rs761908561 Chr.17: 82726970-82726970 on Build GRCh38
rs753047389 Chr.17: 82726802-82726802 on Build GRCh38
rs754005412 Chr.17: 82726887-82726887 on Build GRCh38
rs111958189 Chr.17: 82726855-82726855 on Build GRCh38
rs13955 Chr.17: 82726978-82726978 on Build GRCh38
rs149354070 Chr.17: 82726940-82726940 on Build GRCh38
rs758772179 Chr.17: 82726815-82726815 on Build GRCh38
rs773166303 Chr.17: 82726959-82726959 on Build GRCh38
rs747842191 Chr.17: 82726949-82726949 on Build GRCh38
rs747506396 Chr.17: 82726832-82726832 on Build GRCh38
rs775362232 Chr.17: 82726985-82726985 on Build GRCh38
rs751860519 Chr.17: 82726926-82726926 on Build GRCh38
rs914565618 Chr.17: 82726823-82726823 on Build GRCh38
rs771358908 Chr.17: 82726833-82726833 on Build GRCh38
rs368519234 Chr.17: 82726865-82726865 on Build GRCh38
rs778487608 Chr.17: 82726947-82726947 on Build GRCh38
rs142718764 Chr.17: 82726908-82726908 on Build GRCh38
rs987460815 Chr.17: 82726968-82726968 on Build GRCh38
rs774541187 Chr.17: 82726943-82726943 on Build GRCh38
rs955013102 Chr.17: 82726890-82726890 on Build GRCh38
rs374284375 Chr.17: 82726937-82726937 on Build GRCh38
rs771793858 Chr.17: 82726958-82726958 on Build GRCh38
rs990355272 Chr.17: 82726932-82726932 on Build GRCh38
rs766372633 Chr.17: 82726886-82726886 on Build GRCh38
rs764443615 Chr.17: 82726919-82726919 on Build GRCh38
rs759678845 Chr.17: 82726947-82726967 on Build GRCh38
rs759632794 Chr.17: 82726892-82726892 on Build GRCh38
rs763247815 Chr.17: 82726990-82726990 on Build GRCh38

View Full Product Details